Product Image
Applied Biosystems™
25-bp DNA Ladder
25-bp DNA Ladder is designed for the sizing of double-stranded DNA in the range of 25 to 2,500 bp. ItRead more
Have Questions?
Catalog NumberQuantity
93134350 μg
Catalog number 931343
Price (KRW)
191,000
Each
Quantity:
50 μg
Price (KRW)
191,000
Each
25-bp DNA Ladder is designed for the sizing of double-stranded DNA in the range of 25 to 2,500 bp. It consists of 13 individual chromatography-purified DNA fragments and has reference bands at 2000, 150, and 25 bp for easy orientation.
25-bp DNA Ladder is ideal for separation on 1-4% agarose gels.
Sharp, clear bands—chromatography purified fragments for consistent and reliable results
Designed with Applied Biosystems microarrays in mind—bands were designed to work with multiple Applied Biosystems microarray platforms including CytoScan, Axiom, and OncoScan platforms
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration6X
Gel CompatibilityAgarose Gels, E-Gel™ 48, E-Gel™ 96, TBE Gels, TBE-Urea Gels
Quantity50 μg
Ready to LoadNo
Shipping ConditionDry Ice
Gel TypeAgarose
Product TypeDNA Ladder
Size Range25 to 2500 bp
Unit SizeEach
Contents & Storage
• 1 x 0.5 mL 25 bp DNA Ladder, store at -20°C
• 1 x 0.5 mL 6X Orange DNA Loading Dye, store at -20°C