Fluoro-Max™ Fluorescent Carboxylate-Modified Particles
Thermo Scientific™

Fluoro-Max™ Fluorescent Carboxylate-Modified Particles

Lower detection limits of fluorescent assays by limiting background interference from relatively short-lived matrix fluorescence.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric) MeanQuantity
934703500103500.1 μm100 mL
93470350010150
also known as MCG-93470350010150-EACH
0.1 μm5 mL
93470350011150
also known as MCG-93470350011150-EACH
0.1 μm1 mL
93470520010150
also known as MCG-93470520010150-EACH
0.2 μm5 mL
93470520010350
also known as MCG-93470520010350-EACH
0.2 μm100 mL
93470520011150
also known as MCG-93470520011150-EACH
0.2 μm1 mL
93470720010150
also known as MCG-93470720010150-EACH
0.3 μm5 mL
93470720010350
also known as MCG-93470720010350-EACH
0.3 μm100 mL
93470720011150
also known as MCG-93470720011150-EACH
0.3 μm1 mL
S93470.1, 0.2, 0.3 μm3 x 1 mL
Catalog number 93470350010350
Price (EUR)
-
Diameter (Metric) Mean:
0.1 μm
Quantity:
100 mL
Lower detection limits of fluorescent assays by limiting background interference from relatively short-lived matrix fluorescence with Thermo Scientific™ Fluoro-Max Fluorescent Carboxylate-Modified Particles. Made by dyeing Thermo Scientific™ OptiLink™ carboxylate-modified particles with europium chelate, particles have been specifically designed for membrane or automated fluorometric-based applications. With an extremely broad Stokes shift, Fluoro-Max europium chelate particles help prevent nonspecific fluorescence interference.
Particles are made from carboxylate-modified polystyrene particles containing encapsulated europium chelate dye. They are available in standard 0.1, 0.2 or 0.3μm diameters. When these particles excite at 333nm and emit at 613nm, they do so with a very long lifetime of approximately 0.5 milliseconds, about 10,000 to 100,000 times the lifetime of most fluorophores. The extended lifetime of europium chelate particles allows them to be used in a wide range of time-resolved fluorescence applications.
  • Particles are internally dyed to ensure maximum color brilliance and color saturation, resulting in greater sensitivity, enhanced readability of the assay and elimination of problems associated with enzyme-based assays.
  • Encapsulated dye prevents leaching in aqueous media and provides optimum surface immunoreactivity.
  • Dyeing process causes little or no inhibition of adsorption or covalent couping of immunoglobulin G, resulting in superior binding of protein to the particles.
  • Suitable for a variety of applications such as clinical diagnostics, immuno/histological studies and research.

Recommended for:

Clinical diagnostics, molecular biology and lateral flow, turbidimetric and slide aggulation assays

Specifications
Additive0.05% Sodium Azide
Binding PropertyCarboxylate-modified
Concentration1% solids
ContentsFluorescent red dyed polystyrene microspheres in water
Diameter (Metric) Mean0.1 μm
Diameter (Metric) Nominal0.1 μm
Dye TypeEuropium Chelate
IncludesPackage Insert Sheet containing Certicate of Analysis
Parking Area50 A²/group
Particle Density1.06g/cm³
Quantity100 mL
StabilityProduct stable for a minimum of 36 months
Surface ModificationCarboxylate-Modified
Uniformity<5% CV
ColorWhite
Diameter (Metric)0.1 μm
MaterialPolystyrene or Polystyrene with Coploymer Grafted Surface
Product TypeFluroescent Carboxylate-Modified Particle
Unit SizeEach
Contents & Storage
Unless otherwise stated, refrigerate (2° to 8°C) product when not in use but do not freeze. Shipment at ambient temperature. Store upright and keep bottle tightly sealed. Mix product with gentle inversion by hand or vortex mixer.