Finntip™ Non-Filtered 384 Format Pipette Tips  20 μL
Thermo Scientific™
Finntip™ Non-Filtered 384 Format Pipette Tips 20 μL
Ensure accurate, precise pipetting. Finntip™ Finntip 20 Pipette Tips are designed using our knowledge of liquid handling, with uniformly smooth, hydrophobic surfaces that prevent liquid retention.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호멸균Fit for Purpose형식
9400610Non-sterileCertified QualityRack
9400620Non-sterileCertified QualityBulk
9400613SterileSterile & PCR ReadyRack
카탈로그 번호 9400610
제품 가격(KRW)
750,000
Case of 3840
멸균:
Non-sterile
Fit for Purpose:
Certified Quality
형식:
Rack
제품 가격(KRW)
750,000
Case of 3840
Accurate, precise pipetting with 384-format volume 20 μL. Finntip™ Finntip 20 Pipette Tips are designed using our knowledge of liquid handling, with uniformly smooth, hydrophobic surfaces that prevent liquid retention.
Key Features:
  • Smooth, uniform inner surface of the tips prevents liquid retention
  • Available as Sterile or Non-sterile option
  • Packaging options: racked, bulk bag or reload tower
  • Sterile tips are supplied in vacuum sealed, sterilized rack
Proven quality
  • Sterile options certified free of RNase, DNase, DNA, PCR inhibitors, ATP and endotoxin contamination
Immediate traceability
  • Each tip rack has ink-jetted lot and volume information; the convenient color-coded racks make it easy to match the compatible Finnpipette with the corresponding Finntip pipette tip rack insert
  • Color-coding visible in rack and reload packaging options
Note:
Availability may vary by country.
사양
설명Rack
길이(영국식 단위)1.3 in.
길이(미터법)3.3cm
모델Finntip 20
색상Purple
색상 코드Purple
필터링됨Non-filtered
Fit for PurposeCertified Quality
용도 (장비)Thermo Scientific Finnpipette 20
형식Rack
제품라인FinnTip
멸균Non-sterile
부피(미터법)20 μL
Unit SizeCase of 3840