Especificações
Boiling Point187.6°C (1013 hPa)
CAS57-55-6
Chemical Name or Material(±)-1,2-Propanediol
ColorColorless
Melting Point-59°C
Molecular FormulaCH3CH(OH)CH2OH
SynonymPropylene glycol, (±)-1,2-Propylene glycol, 1,2-Dihydroxypropane, 1,2-Propanediol, (±)-1,2-Dihydroxy propane
GradeMulti-Compendial
Molecular Weight (g/mol)76.1
Percent Purity≥99.5%
Quantity500 mL
Unit Size500 mL