Finntip™ Flex Filtered Pipette Tips
Finntip™ Flex Filtered Pipette Tips
Finntip™ Flex Filtered Pipette Tips
Finntip™ Flex Filtered Pipette Tips
Finntip™ Flex Filtered Pipette Tips
Thermo Scientific™

Finntip™ Flex Filtered Pipette Tips

Minimize the risk of cross-contamination with these flex filtered pipette tips that combine the ergonomics of a standard pipette tip with the safety of a filter tip.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
94056520100 μL
9405698010 μL
9405651030 μL
94056380200 μL
94056580300 μL
940567101000 μL
940568101200 μL
9405697010 mL
Catalog number 94056520
Price (USD)
192.00
Online exclusive
256.00
Save 64.00
Case of 960
Volume (Metric):
100 μL
Price (USD)
192.00
Online exclusive
256.00
Save 64.00
Case of 960

Minimize the risk of cross-contamination with Thermo Scientific™ Finntip™ Flex Filter Pipette Tips, which combine the ergonomics of a standard Finntip Flex pipette tip and the safety of a filter tip. Offered sterile, and free from DNA, DNase and RNase. Ideal for PCR and pipetting of radioactive and infectious samples. The filters, made of an inert nonsealing polyethylene matrix, are effective at eliminating carryover contamination from liquids and aerosols between pipetter and tip.

Key Features:

  • Excellent for single and multichannel pipetting
  • Smooth, uniform inner surface of the tips prevents liquid retention
  • Filtered Finntip Flex pipette tips are ideal for sensitive assays and with critical samples to prevent cross-contaminations
  • Inert and non-sealing polyethylene filter
  • Tips are supplied in vacuum sealed, sterilized, 10 x 96 rack (10 mL Extended in bag)

Softer Finntip Flex Tip

  • The softer tip collar reduces the amount of force needed to attach and eject tips, significantly lowering the risk of repetitive stress injury (RSI)
  • It also ensures a better seal with the pipetter module, ensuring higher accuracy and precision

Proven quality

  • Certified free of RNase, DNase, DNA, PCR inhibitors, ATP and endotoxin contamination (excluding 10 mL Extended)

Versatile Tip Range

  • Available in volumes: 10, 30, 100, 200, 300, 1000, 1200 μL and 10mL Extended (bottle tip)

Immediate traceability

  • Each tip rack has ink-jetted lot and volume information; the convenient color-coded racks make it easy to match the compatible Finnpipette with the corresponding Finntip Flex pipette tip rack insert

Note:

Availability may vary by country.

Specifications
DescriptionFinntip Flex Filter 100 μL, sterile
Length (Metric)6 cm
Color CodeYellow
FilteredFiltered
Fit for PurposeSterile & PCR Ready
For Use With (Equipment)Thermo Scientific Finnpipette
FormatRack
Product LineFinnTip Flex
SterilitySterile
Volume (Metric)100 μL
Unit SizeCase of 960

Frequently asked questions (FAQs)

Can I autoclave Finntip Flex Filtered Pipette Tips and the rack they come in?

No, the Finntip Flex Filtered Pipette Tips cannot be autoclaved. However, the empty racks can be autoclaved. We recommend using the following autoclave conditions for PolyPropylene material: 121 degrees C, 1 bar for 20 minutes.