FluidEase™ Pro ClipTip™ Electronic Pipette Charging Stand and Extension Kit
Thermo Scientific™
FluidEase™ Pro ClipTip™ Electronic Pipette Charging Stand and Extension Kit
The Thermo Scientific™ FluidEase™ Pro ClipTip™ electronic pipette charging stand enables simultaneous wireless charging of up to four pipettes and provides dedicated resting positions for short workflow pauses, keeping electronic pipettes powered, accessible, and organized in shared laboratories.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With
9420530FluidEase Pipette Charging Stand
9420520FluidEase Pipettes
Catalog number 9420530
Price (USD)
99.00
Pack of 1
For Use With:
FluidEase Pipette Charging Stand
Price (USD)
99.00
Pack of 1
The Thermo Scientific™ FluidEase™ Pro ClipTip™ electronic pipette charging stand enables simultaneous wireless charging of up to four electronic pipettes without individual cables. Dedicated resting positions allow users to temporarily place pipettes with tips attached and liquid inside during short workflow interruptions. A modular extension design supports scalable charging capacity for shared laboratory environments.
Features:
  • Electronic Pipette Charging Stand for Shared Laboratories: Designed specifically for Thermo Scientific™ FluidEase™ Pro ClipTip™ electronic pipettes used in single- and multichannel workflows.
  • Simultaneous Charging of Up to Four Pipettes: Supports wireless charging of up to four electronic pipettes at the same time, reducing cable clutter and bench complexity.
  • Dedicated Charging and Resting Positions: Includes active charging positions and separate temporary resting positions for safely placing pipettes during short workflow pauses.
  • Temporary Resting with Tips and Liquid Inside: Allows users to briefly rest pipettes with tips attached and liquid retained when reaching for tubes, reagents, or plates.
  • Modular and Scalable Design: Extension kits enable charging capacity to scale from one to four pipettes as laboratory needs grow.
  • Organized Bench Footprint: Keeps electronic pipettes visible, accessible, and securely positioned to support efficient shared-lab operation.
Specifications
Quantity1 Ea.
ColorWhite
For Use WithFluidEase Pipette Charging Stand
TypeCharging Stand Extension Kit
Unit SizePack of 1