Hollow Cathode Lamps for iCE™ 3000 Series Atomic Absorption Spectrometers
Actual product may vary
Thermo Scientific™
Hollow Cathode Lamps for iCE™ 3000 Series Atomic Absorption Spectrometers
For maximum flexibility from your Thermo Scientific™ iCE™ 3000 Series AAS system, we offer a huge range of pre-coded and un-coded Hollow Cathode Lamps.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeLamp
942339331131Hollow Cathode Lamp Data CodedCr/Cu Data Coded
942339020051Hollow Cathode Lamps, UncodedBoron Uncoded
942339020031Hollow Cathode Lamps, UncodedLithium Uncoded
942339020111Hollow Cathode Lamps, UncodedSodium Uncoded
942339020041Hollow Cathode Lamps, UncodedBeryllium Uncoded
942339020121Hollow Cathode Lamps, UncodedMagnesium Uncoded
94233902011VHollow Cathode Lamps, UncodedSodium (Na) Uncoded
942339020131Hollow Cathode Lamps, UncodedAluminum Uncoded
942339020141Hollow Cathode Lamps, UncodedSilicon Uncoded
94233902012VHollow Cathode Lamps, UncodedMagnesium (Mg) Uncoded
94233902013VHollow Cathode Lamps, UncodedAluminum (Al) Uncoded
942339020151Hollow Cathode Lamps, UncodedPhosphorus Uncoded
942339020201Hollow Cathode Lamps, UncodedCalcium Uncoded
942339020191Hollow Cathode Lamps, UncodedPotassium Uncoded
94233902019VHollow Cathode Lamps, UncodedPotassium (K) Uncoded
94233902020VHollow Cathode Lamps, UncodedCalcium (Ca) Uncoded
942339020231Hollow Cathode Lamps, UncodedVanadium Uncoded
942339020211Hollow Cathode Lamps, UncodedScandium Uncoded
942339020221Hollow Cathode Lamps, UncodedTitanium Uncoded
942339020241Hollow Cathode Lamps, UncodedChromium Uncoded
942339020251Hollow Cathode Lamps, UncodedManganese Uncoded
94233902024VHollow Cathode Lamps, UncodedChromium (Cr) Uncoded
94233902025VHollow Cathode Lamps, UncodedManganese (Mn) Uncoded
94233902026VHollow Cathode Lamps, UncodedIron (Fe) Uncoded
942339020271Hollow Cathode Lamps, UncodedCobalt Uncoded
942339020281Hollow Cathode Lamps, UncodedNickel Uncoded
94233902027VHollow Cathode Lamps, UncodedCobalt (Co) Uncoded
94233902028VHollow Cathode Lamps, UncodedNickel (Ni) Uncoded
942339020291Hollow Cathode Lamps, UncodedCopper Uncoded
94233902029VHollow Cathode Lamps, UncodedCopper (Cu) Uncoded
942339020301Hollow Cathode Lamps, UncodedZinc Uncoded
942339020321Hollow Cathode Lamps, UncodedGermanium Uncoded
94233902030VHollow Cathode Lamps, UncodedZinc (Zn) Uncoded
942339020311Hollow Cathode Lamps, UncodedGallium Uncoded
94233902033VHollow Cathode Lamps, UncodedArsenic (As) Uncoded
942339020331Hollow Cathode Lamps, UncodedArsenic Uncoded
942339020371Hollow Cathode Lamps, UncodedRubidium Uncoded
942339020341Hollow Cathode Lamps, UncodedSelenium Uncoded
94233902034VHollow Cathode Lamps, UncodedSelenium (Se) Uncoded
942339020391Hollow Cathode Lamps, UncodedYttrium Uncoded
942339020381Hollow Cathode Lamps, UncodedStrontium Uncoded
942339020401Hollow Cathode Lamps, UncodedZirconium Uncoded
942339020451Hollow Cathode Lamps, UncodedRhodium Uncoded
942339020411Hollow Cathode Lamps, UncodedNiobium Uncoded
94233902042VHollow Cathode Lamps, UncodedMolybdenum (Mo) Uncoded
942339020441Hollow Cathode Lamps, UncodedRuthenium Uncoded
942339020421Hollow Cathode Lamps, UncodedMolybdenum Uncoded
942339020471Hollow Cathode Lamps, UncodedSilver Uncoded
94233902047VHollow Cathode Lamps, UncodedSilver (Ag) Uncoded
942339020481Hollow Cathode Lamps, UncodedCadmium Uncoded
94233902048VHollow Cathode Lamps, UncodedCadmium (Cd)Uncoded
942339020511Hollow Cathode Lamps, UncodedAntimony Uncoded
942339020491Hollow Cathode Lamps, UncodedIndium Uncoded
942339020501Hollow Cathode Lamps, UncodedTin Uncoded
94233902051VHollow Cathode Lamps, UncodedAntimony (Sb) Uncoded
94233902050VHollow Cathode Lamps, UncodedTin (Sn) Uncoded
942339020521Hollow Cathode Lamps, UncodedTellurium Uncoded
942339020461Hollow Cathode Lamps, UncodedPalladium Uncoded
942339020561Hollow Cathode Lamps, UncodedBarium Uncoded
942339020571Hollow Cathode Lamps, UncodedLanthanum Uncoded
942339020551Hollow Cathode Lamps, UncodedCaesium Uncoded
942339020591Hollow Cathode Lamps, UncodedPraseodymium Uncoded
942339020601Hollow Cathode Lamps, UncodedNeodymium Uncoded
942339020631Hollow Cathode Lamps, UncodedEuropium Uncoded
942339020641Hollow Cathode Lamps, UncodedGadolinium Uncoded
942339020621Hollow Cathode Lamps, UncodedSamarium Uncoded
942339020651Hollow Cathode Lamps, UncodedTerbium Uncoded
942339020671Hollow Cathode Lamps, UncodedHolmium Uncoded
942339020661Hollow Cathode Lamps, UncodedDysprosium Uncoded
942339020681Hollow Cathode Lamps, UncodedErbium Uncoded
942339020721Hollow Cathode Lamps, UncodedHafnium Uncoded
942339020711Hollow Cathode Lamps, UncodedLutetium Uncoded
942339020731Hollow Cathode Lamps, UncodedTantalum Uncoded
942339020701Hollow Cathode Lamps, UncodedYtterbium Uncoded
942339020691Hollow Cathode Lamps, UncodedThulium Uncoded
942339020741Hollow Cathode Lamps, UncodedTungsten Uncoded
942339020751Hollow Cathode Lamps, UncodedRhenium (Re) Uncoded
942339020761Hollow Cathode Lamps, UncodedOsmium Uncoded
942339020781Hollow Cathode Lamps, UncodedPlatinum Uncoded
942339020791Hollow Cathode Lamps, UncodedGold Uncoded
942339020771Hollow Cathode Lamps, UncodedIridium Uncoded
94233902079VHollow Cathode Lamps, UncodedGold (Au) Uncoded
942339020801Hollow Cathode Lamps, UncodedMercury Uncoded
94233902080VHollow Cathode Lamps, UncodedMercury (Hg) Uncoded
942339020811Hollow Cathode Lamps, UncodedThallium Uncoded
942339020821Hollow Cathode Lamps, UncodedLead Uncoded
942339020831Hollow Cathode Lamps, UncodedBismuth Uncoded
94233902082VHollow Cathode Lamps, UncodedLead (Pb) Uncoded
94233902083VHollow Cathode Lamps, UncodedBismuth (Bi) Uncoded
942339030011Hollow Cathode Lamp Data CodedDeuterium Data Coded
942339030041Hollow Cathode Lamp Data CodedBeryllium Data Coded
942339030051Hollow Cathode Lamp Data CodedBoron Data Coded
942339030031Hollow Cathode Lamp Data CodedLithium Data Coded
942339030121Hollow Cathode Lamp Data CodedMagnesium Data Coded
942339030141Hollow Cathode Lamp Data CodedSilicon Data Coded
942339030131Hollow Cathode Lamp Data CodedAluminum Data Coded
942339030111Hollow Cathode Lamp Data CodedSodium Data Coded
942339030151Hollow Cathode Lamp Data CodedPhosphorus Data Coded
942339030191Hollow Cathode Lamp Data CodedPotassium Data Coded
942339030201Hollow Cathode Lamp Data CodedCalcium Data Coded
Catalog number 942339331131
Price (BRL)
-
Type:
Hollow Cathode Lamp Data Coded
Lamp:
Cr/Cu Data Coded
For maximum flexibility from your Thermo Scientific™ iCE™ 3000 Series Atomic Absorption spectrometer, a huge range of pre-coded and un-coded Thermo Scientific™ Hollow Cathode Lamps are available, to handle your chosen elements for analysis. Whether you require a single dedicated element or a multi-element cathode lamp, there is a one to match your needs, whether coded or un-coded.
For maximum flexibility from your Thermo Scientific™ iCE™ 3000 Series Atomic Absorption spectrometer, a huge range of pre-coded and un-coded Thermo Scientific Hollow Cathode Lamps are available, to handle your chosen elements for analysis. Whether you require a single dedicated element or a multi-element cathode lamp, there is a one to match your needs, whether coded or un-coded.
Specifications
LampCr/Cu Data Coded
For Use With (Equipment)iCE 3000 Series Atomic Absorption Spectrometers
TypeHollow Cathode Lamp Data Coded
Unit SizeEach