Sample and Reagent Cups for iCE™ 3000 Series AA systems
Actual product may vary
Thermo Scientific™
Sample and Reagent Cups for iCE™ 3000 Series AA systems
Ensure consistent sample presentation and complete sample pick-up with sample and reagent cups for Thermo Scientific™ iCE™ 3000 Series AA systems.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
942339380051PTFE Sample Cups
942339380031Polypropylene Sample Cups
942339380061Reduced Volume Sample Cups
942339380021Polypropylene Reagent Cups
942339381261Spare Sampling Capillary Tips
Catalog number 942339380051
Price (BRL)
-
Type:
PTFE Sample Cups
Use Thermo Scientific™ Sample and Reagent Cups with Thermo Scientific™ GFS33 and GFS35(Z) Graphite Furnace Autosamplers to ensure consistent sample presentation and complete sample pick-up. Select from polypropylene sample and reagent cups, PTFE sample cups, and reduced volume sample cups.
The perfect choice for the Thermo Scientific™ GFS33 and GFS35(Z) Graphite Furnace Autosamplers, when used with our iCE 3000 Series Atomic Absorption spectrometers; these Thermo Scientific Sample and Reagent Cups for Thermo Scientific™ iCE™ 3000 Series AA systems ensure consistent sample presentation and complete sample pick-up. Select from polypropylene sample and reagent cups, PTFE sample cups, and reduced volume sample cups, according to your needs.
Specifications
Quantity20/Pk.
For Use With (Equipment)GFS33 and GFS35(Z) Graphite Furnace Autosamplers
TypePTFE Sample Cups
Unit SizePack of 20