Product Image
Thermo Scientific™
Graphite Contacts Cones for GFS33 or GFS35 Furnaces
Easy replacement Thermo Scientific™ Graphite Furnace Cones keep your Thermo Scientific™ GFS33 or GFS35 furnaces for our AA instruments in top operational condition.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeFor Use With (Equipment)
942339395011Graphite Contacts ConesGFS33 or GFS35 Furnaces
942339395161Graphite Contacts Cones (Pair)GF95 Z Furnace
Catalog number 942339395011
Price (MXN)
-
Type:
Graphite Contacts Cones
For Use With (Equipment):
GFS33 or GFS35 Furnaces
Designed for maximum ease of use, the high-performance combined Thermo Scientific™ Graphite Furnace and Autosampler modules (GFS33 and GFS35) for furnace Atomic Absorption analysis use a simple lever system to open and close the furnace head, for easy replacement of Thermo Scientific Graphite Contact Cones for Thermo Scientific GFS33 or GFS35 furnaces.
Specifications
For Use With (Equipment)GFS33 or GFS35 Furnaces
TypeGraphite Contacts Cones
Unit SizeEach