Product Image
Thermo Scientific™
Potassium Dichromate Solution Standard, Set of 2
Potassium Dichromate Solution Standards are NIST-traceable for qualifying instrument performance.
Catalog NumberQuantity
9423UV95100ESet of 2
9423UV95200ESet of 6
Catalog number 9423UV95100E
Price (JPY)
-
Quantity:
Set of 2
Thermo Scientific™ Potassium Dichromate Solution Standards are NIST-traceable for qualifying instrument performance. Qualify your spectrophotometer's performance with NIST-traceable Thermo Scientific Potassium Dichromate Solution Standards for photometric accuracy.
The Potassium Dichromate Solution Standard is available in a set of two or six standards. Both standard sets are:
  • Supplied in 0.001M perchloric acid in permanently sealed quartz cells
  • Compatible with any cell holder that accommodates 10mm pathlength rectangular cells
  • Traceable to NIST™ standards and supplied with a calibration certificate
  • Calibrated for use with instruments with a SBW of ≤1.6nm, including the Thermo Scientific Evolution 60, Evolution 300 and Evolution 600
Set of Two Standards
  • Intended for testing photometric accuracy at 430nm as required by the European Pharmacopoeia Section 2.2.25, revised 2005
  • Includes a 60mg/L standard and corresponding blank
Set of Six Standards
  • Designed to check linearity of the instrument in the UV region
  • Calibrated at 235, 257, 313 and 350nm
  • Includes standards at 0, 20, 40, 60, 80 and 100mg/L concentrations
Specifications
CalibrationNIST Traceable
For Use With (Equipment)Instruments with a SBW of ≤1.6nm; Evolution 60; Evolution 300; Evolution 600
Includes0 and 60mg/L standards
QuantitySet of 2
Wavelength Range235 to 350 nm
Unit SizeEach