Labtip Pipette Tips
Thermo Scientific™

Labtip Pipette Tips

Basic pipette tips for standard liquid transfer needs.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoVolume (Metric)FormatPackaging
94300130200 μLRack10 x 96/Tray
943002101000 μLBulk10,000 tips/bag
94300120200 μLBulk1000 tips/bag
94300320300 μLBulk1000 tips/bag
Número do catálogo 94300130
Preço (BRL)
-
Volume (Metric):
200 μL
Format:
Rack
Packaging:
10 x 96/Tray
Thermo Scientific™ Labtip standard pipette tips are made from 100% virgin polypropylene. These tips are available in 10, 200, 300 and 1000 μL sizes. Available in clear (10 μL and 300 μL), yellow (200 μL), blue (1000 μL), these tips for general lab use across countless liquid transfer applications.
Labtip pipette tips provide:
  • The most common sizes used in a lab: 10, 200, 300 and 1000 μL
  • Non-Filter essential tips for general laboratory usage
  • Non-sterile packaging in both Racked and Bulk options
Packaging Options:
  • Racked, nonsterile - 96 tips/ tray, 10 trays/pack
  • Bulk - 1,000 tips/ bag
  • Bulk - 25,000 tips/bag
Especificações
DescriptionLabtip 200
Length (Metric)4.9cm
Packaging10 x 96/Tray
Quantity4 Trays/Pk.
ColorYellow
FilteredNon-filtered
For Use With (Equipment)Thermo Scientific Finnpipette
FormatRack
MaterialVirgin Polypropylene
Product LineLabTip
SterilityNon-sterile
Volume (Metric)200 μL
Unit SizeCase of 3840