ClipTip™ Pipette Tip Racks
Actual product may vary
ClipTip™ Pipette Tip Racks
ClipTip™ Pipette Tip Racks
ClipTip™ Pipette Tip Racks
ClipTip™ Pipette Tip Racks
ClipTip™ Pipette Tip Racks
ClipTip™ Pipette Tip Racks
ClipTip™ Pipette Tip Racks
ClipTip™ Pipette Tip Racks
ClipTip™ Pipette Tip Racks
ClipTip™ Pipette Tip Racks
ClipTip™ Pipette Tip Racks
ClipTip™ Pipette Tip Racks
ClipTip™ Pipette Tip Racks
ClipTip™ Pipette Tip Racks
Thermo Scientific™
ClipTip™ Pipette Tip Racks
Utilize Thermo Scientific™ ClipTip™ Pipette Tip Racks, available in three sizes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
94410819ClipTip Empty Rack, Large
94410219ClipTip Empty Rack, Small
94410619ClipTip Empty Rack
94410519ClipTip Empty Rack, Medium
Catalog number 94410819
Price (MXN)
-
Description:
ClipTip Empty Rack, Large
Utilize Thermo Scientific™ ClipTip™ Pipette Tip Racks, available in three sizes.
  • Small Rack: For use with ClipTip 20 and 50 Pipette Tips
  • Medium Rack: For use with ClipTip 200 and 300 Pipette Tips
  • Large Rack: For use with ClipTip 1000 Pipette Tips
Ordering Information:
Racks are supplied without tips and inserts
Specifications
DescriptionClipTip Empty Rack, Large
ModelClipTip Empty Rack
For Use With (Equipment)ClipTip 1000 Pipette Tips (94420718)
FormatRack
Product LineClipTip
Unit SizeCase of 8