ClipTip™ Filtered Pipette Tips
Thermo Scientific™

ClipTip™ Filtered Pipette Tips

Ensure a secure seal for every sample. A unique, innovative interlocking technology ensures a complete seal on every channel with minimal tip attachment and ejection force.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)FormatTip Style
9442004312.5 μLRackStandard
9442021820 μLReload DeckLow Retention
9442025850 μLReload DeckLow Retention
94420318200 μLReload DeckLow Retention
94420518300 μLReload DeckLow Retention
944207181000 μLReload DeckLow Retention
944208131250 μLRackLow Retention
944208181250 μLReload DeckLow Retention
9442021320 μLRackLow Retention
9442025350 μLRackLow Retention
94420313200 μLRackLow Retention
94420513300 μLRackLow Retention
944207131000 μLRackLow Retention
9442006312.5 μLRackExtended Length
94420613300 μLRackLow Retention, Extended Length
Catalog number 94420043
Price (USD)
123.30
Online Exclusive
137.00
Save 13.70
Case of 960
Volume (Metric):
12.5 μL
Format:
Rack
Tip Style:
Standard
Price (USD)
123.30
Online Exclusive
137.00
Save 13.70
Case of 960
Ensure a secure seal for every sample. Thermo Scientific™ ClipTip™ Pipette Tips provide a secure seal for every sample with a unique and innovative interlocking technology that ensures a complete seal on every channel with minimal tip attachment and ejection force. Achieve newfound confidence knowing that once attached, your tips are locked firmly in place, and will not loosen or fall off regardless of application pressure. The innovative three interlocking clip design ensures the tip is held securely on the F1-ClipTip and E1-ClipTip pipettes until — and only until — it is released.

Key Features:

  • ClipTip pipette tips feature 'clips' that lock tips in place and form a complete seal
  • Each time ClipTip pipette tips are on, they are sealed and will not loosen regardless of application pressure
  • ClipTip pipette tips are clipped securely on the F1-ClipTip and E1-ClipTip pipettes with a light touch eliminating the frustration of banging tips on your pipette
  • Filtered ClipTip pipette tips are ideal for sensitive assays, such as PCR, and with critical samples to prevent cross-contaminations

Low retention

  • Low retention technology minimizes liquid retention, and enable maximium sample recovery. Available in ClipTip 20-1250 96-format tips.


Proven quality

  • Certified free of RNase, DNase, DNA, PCR inhibitors, ATP and endotoxin contamination


Environmentally responsible

  • ClipTip reload system uses recycled components and 30% less plastic


Versatile Tip Range

  • 96-format volumes: 12.5, 20, 50, 200, 300, 1000 and 1250μL
  • 384-format volumes: 12.5, 30 and 125μL
  • Extended length tips available for volumes 12.5 and 300 μl allowing for better reach with specific labware consumables


Immediate traceability

  • Each tip insert has ink-jetted lot and volume information; the convenient color key makes it easy to match the color code of the F1-ClipTip and E1-ClipTip pipette with the corresponding ClipTip pipette tip rack insert


Note:

Availability may vary by country.

Specifications
DescriptionClipTip 12.5, Filtered, Sterile, Rack
Length (English)1.2 in.
Length (Metric)3.0cm
Packaging10 x 96 tips/rack
Quantity960 tips
AutoclavableNot Autoclavable
Color CodePink
FilteredFiltered
Fit for PurposeSterile & PCR Ready
For Use With (Equipment)Thermo Scientific E1-ClipTip, Thermo Scientific F1-ClipTip
FormatRack
Product LineClipTip
SterilitySterile
Tip StyleStandard
Volume (Metric)12.5 μL
Unit SizeCase of 960