AcroMetrix™ HCV Controls (RUO)
Product Images
Thermo Scientific™
AcroMetrix™ HCV Controls (RUO)
Identify random or systematic errors in nucleic acid test procedures for hepatitis C viral RNA (HCV RNA) with these controls, which are for Research Use Only.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLevelPackaging
950201Low5 x 1mL
950202Mid5 x 1mL
2 Options
Catalog number 950201
Price (USD)/ Each
-
Request A Quote
Level:
Low
Packaging:
5 x 1mL
Monitor day-to-day test variation, lot-to-lot performance of test kits, and operator variation with the Thermo Scientific™ AcroMetrix™ HCV Controls (RUO). These controls identify random or systematic errors in nucleic acid test procedures for the quantitative and qualitative determination of hepatitis C viral RNA (HCV RNA) and can assist in identifying increases in random or systematic error. The AcroMetrix HCV Controls (RUO) are not intended for use with assays used to screen the blood supply. The controls are for Research Use Only and are not for use in diagnostic procedures.
For Research Use Only. Not for use in diagnostic procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionAcroMetrix NAC HCV Low Control
Detectable AnalytesHCV
TypeHCV Control
LevelLow
Packaging5 x 1mL
Quantity5 x 1 mL
Unit SizeEach