Microlite™ White Microtiter™ Plates
Microlite™ White Microtiter™ Plates
Actual product may vary
Thermo Scientific™
Microlite™ White Microtiter™ Plates
Experience consistent binding, high reflectivity, and low crosstalk for use in luminescent immunoassays and related techniques using Microlite Microtiter Plates.
Have Questions?
Catalog NumberProduct Type
9503100Strip Removal Tool
Catalog number 9503100
Price (USD)
-
Request A Quote
Product Type:
Strip Removal Tool
Experience consistent binding, high reflectivity, and low crosstalk for use in luminescent immunoassays and related techniques using Thermo Scientific™ Microlite White Microtiter Plates.
Available in plates or 12-well breakable strips.
  • Polystyrene
  • Choice of two surfaces, each performance assayed using an IgG binding assay
SURFACES
  • Microlite 1+ - Hydrophobic surface for binding of biomolecules with hydrophilic domains
  • Microlite 2+ - Slightly hydrophilic surface for increased binding of biomolecules with hydrophilic/hydrophobic character such as many proteins
Note:
Other configurations are available in Microlite 1 and 2 formulations.
Specifications
Binding PropertySlightly Hydrophilic
Binding TypeAdsorption
Detection MethodChemiluminescence
Plate BlockingNone
Product TypeStrip Removal Tool
Target MoleculeAntibodies (Glycoproteins) and Proteins
ColorWhite
MaterialPolystyrene
SterilityNon-sterile
Unit Size-