KingFisher 96 Deep-well, (for KF Flex/Duo Prime only) - FAQs
KingFisher 96 Deep-well, (for KF Flex/Duo Prime only) - FAQs
View additional product information for KingFisher 96 Deep-well Plates - FAQs (95040455)
6 product FAQs found
Which deep-well plate would be a suitable alternative for the KingFisher 96 deep-well plate (Cat. No. 95040450)?
The following catalog numbers would be a suitable alternative: 95040460, A48305, 95040455 and A48424.
Do you offer an alternative for the discontinued Pharma MagMAX Express-96 Deep-Well Plates (Cat. No. A31540)?
Yes, we recommend using KingFisher 96 deep-well plate (Cat. No. 95040450, 95040460, or 95040455).
What is the total well volume for a KingFisher 96 deep-well plate?
The total well volume for a KingFisher 96 deep-well plate is 2 mL.
Are KingFisher Plastics for 96-deep well format autoclavable?
No, KingFisher Plastics for 96-deep well format are not autoclavable. They won't melt, but they will get deformed.
Can I reuse the Kingfisher Plastics for 96 deep-well format?
The Kingfisher Plastics for 96 deep-well format are recommended for one-time use only, and we don't recommend reusing these products.
Are the KingFisher Plastics for 96 deep-well format CE marked?
The KingFisher Plastics for 96 deep-well format are not CE marked.