KingFisher™ Plastics for 24 deep-well format
Thermo Scientific™
KingFisher™ Plastics for 24 deep-well format
Optimize your performance using Thermo Scientific™ KingFisher consumables designed specifically for use with the KingFisher Flex, Duo Prime, and Presto instruments.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeFor Use With (Equipment)
95040480B24 DW plate, barcoded, sterileKingFisher Apex
9700258024 Combi tip combKingFisher Apex
9504047024 Deep-well PlateKF Duo Prime, Flex, Presto
9504048024 Deep-well Plate, SterileKF Duo Prime, Flex, Presto
9700261024 DW tip comb and plateKingFisher Flex and Presto
9700262024 DW tip comb, plate, sterileKingFisher Flex and Presto
970035106-tip comb and 24 DW plateKingFisher Duo Prime
95040470B24 Deep Well plate, barcodedKingFisher Apex
97002610B24 DW tip comb, plate, barcodeKingFisher Apex
97002620B24 DW tip comb, plate, sterileKingFisher Apex
Catalog number 95040480B
Price (MXN)
-
Product Type:
24 DW plate, barcoded, sterile
For Use With (Equipment):
KingFisher Apex
Optimize your performance using Thermo Scientific™ KingFisher Plastics designed specifically for use with the KingFisher Flex, Duo Prime, Presto and Apex instruments. KingFisher instruments utilize disposable plastic plates and tip combs made of polypropylene, which are ideal for magnetic particle processing due to their low binding affinity for biomolecules. Both the yield and quality of the isolated protein or nucleic acid are significantly improved with special plates and tip combs designed for KingFisher systems.
Use With: Thermo Scientific™ KingFisher instruments, including KingFisher Flex, KingFisher Duo Prime, and KingFisher Presto.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
For research use only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)KingFisher Apex
Quantity50/Cs.
Product Type24 DW plate, barcoded, sterile
Unit SizeCase of 50