AcroMetrix™ Wild-Type KRAS FFPE Controls (RUO)
Thermo Scientific™  Related applications: Diagnostic Testing
Assess the performance of the complete KRAS testing workflow, including the isolation, purification, and detection of wild-type KRAS DNA in formalin-fixed paraffin-embedded (FFPE) samples, with the Thermo Scientific™ AcroMetrix™ Wild-Type KRAS FFPE Controls (RUO). These controls have been formulated to mimic FFPE tissues and are designed to monitor all procedural steps in the extraction of human DNA from FFPE samples. The controls are for Research Use Only and are not for use in diagnostic procedures.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
Showing {{showMoreLimit}} of {{commerceData.products.length}} |  Showing {{shownProducts.length}} of {{commerceData.products.length}} | 
Back to top
Description
Assess the performance of the complete KRAS testing workflow, including the isolation, purification, and detection of wild-type KRAS DNA in formalin-fixed paraffin-embedded (FFPE) samples, with the Thermo Scientific™ AcroMetrix™ Wild-Type KRAS FFPE Controls (RUO). These controls have been formulated to mimic FFPE tissues and are designed to monitor all procedural steps in the extraction of human DNA from FFPE samples. The controls are for Research Use Only and are not for use in diagnostic procedures.
For Research Use Only. Not for use in diagnostic procedures.
Documents
Manuals & protocols