AcroMetrix™ EBV CSF Controls (RUO)
Thermo Scientific™  Related applications: Diagnostic Testing
Assess the analytical performance of molecular test procedures, including the DNA extraction step, for the quantitative and qualitative determination of Epstein-Barr Virus (EBV) DNA with the Thermo Scientific™ AcroMetrix™ EBV CSF Controls (RUO). These controls can be used for training and proficiency testing of laboratory personnel, for lot-to-lot comparison of reagent test kits and to evaluate and compare intralaboratory and interlaboratory assessments. The controls are for Research Use Only and are not for use in diagnostic procedures.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
Back to top
Description
Assess the analytical performance of molecular test procedures, including the DNA extraction step, for the quantitative and qualitative determination of Epstein-Barr Virus (EBV) DNA with the Thermo Scientific™ AcroMetrix™ EBV CSF Controls (RUO). These controls can be used for training and proficiency testing of laboratory personnel, for lot-to-lot comparison of reagent test kits and to evaluate and compare intralaboratory and interlaboratory assessments. The controls are for Research Use Only and are not for use in diagnostic procedures.
For Research Use Only. Not for use in diagnostic procedures.
Documents
Manuals & protocols