Product Image
Applied Biosystems™

Axiom™ GeneTitan™ Hybridization Trays, bulk, 96 format

Have Questions?
Número do catálogoQuantity
95235740 each
Número do catálogo 952357
Preço (BRL)
-
Quantity:
40 each
Axiom GeneTitan Hybridization Trays allow for processing of ten Axiom 96-format array plates using the GeneTitan MC instrument. The hybridization trays are required along with GeneTitan barcoded stain trays/covers and scan trays/covers to process arrays on the instrument. The bulk packaging is ideal for high throughput processing of arrays.
Especificações
For Use With (Equipment)Axiom 96 Format Arrays
No. of Samples3840
Quantity40 each
TypeHybridization Tray
Format96-array Plate
Number of Arrays40 arrays
Unit Size40 each
Conteúdo e armazenamento
Room Temperature