Acetic acid, glacial, 99.5-100.05%, USP, Multi-Compendial, GMP Manufactured Product, J.T.Baker™
Avantor™

Acetic acid, glacial, 99.5-100.05%, USP, Multi-Compendial, GMP Manufactured Product, J.T.Baker™

This product is GMP Manufactured and meets JP, BP, USP and EP Requirements.
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
500 mL
2.5 L
15 L
450 lb.
Envase:
Ver todo
Clear glass bottle
Número de catálogo 9526-01
Precio (MXN)
-
Cantidad:
500 mL
Envase:
Clear glass bottle
This product is GMP Manufactured and meets JP, BP, USP and EP Requirements.
This product is GMP Manufactured and meets JP, BP, USP and EP Requirements.
Especificaciones
Punto de ebullición118°C (1013 hPa)
CAS64-19-7
Nombre del producto químico o materialAcetic acid glacial
ColorIncolora
Punto de fusión17°C
Fórmula molecularH3CCOOH
EnvaseClear glass bottle
SinónimoMethanecarboxylic acid, Glacial acetic acid, Vinegar acid, Ethanoic acid
GradoMulti-Compendial/NF
Peso molecular (g/mol)60.05
Porcentaje de pureza99.5-100.05%
Cantidad500 mL
Unit SizeEach