Conductive Automation Tips for Qiagen™ Workstations
Conductive Automation Tips for Qiagen™ Workstations
Conductive Automation Tips for Qiagen™ Workstations
Thermo Scientific™
Conductive Automation Tips for Qiagen™ Workstations
Increase pipetting performance with Thermo Scientific™ Automation Tips for Qiagen™ Universal Pipetting Systems, which undergo a rigorous 15-point quality control program to ensure consistency.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)FilteredFit for Purpose
953-2511100 μLNon-filteredSterile
953-0111000 μLFilteredSterile and PCR Ready
953-2521100 μLNon-filteredCertified Quality
951-252300 μLNon-filteredCertified Quality
951-251300 μLNon-filteredSterile
Catalog number 953-251
Price (USD)
210.60
Online exclusive
234.00
Save 23.40
Case of 960
Volume (Metric):
1100 μL
Filtered:
Non-filtered
Fit for Purpose:
Sterile
Price (USD)
210.60
Online exclusive
234.00
Save 23.40
Case of 960
Increase pipetting performance with Thermo Scientific™ Automation Tips for Qiagen™ Universal Pipetting Systems, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: Qiagen Rosys, Colibri and BioRobot 3000/8000/9600/9604.
Features:
  • Available as sterile and filtered.
  • Sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen.
  • Lot-tested during production on actual workstation for which they are created.
  • Low percentage coefficient of variation (%CV).
Warranty:: A five (5)-year shelf life from the date of manufacture is recommended.
WARNING: Cancer – www.P65Warnings.ca.gov
Specifications
Packaging96 Tips/Tray, 10 Trays/Case
ColorBlack
FilteredNon-filtered
Fit for PurposeSterile
For Use With (Equipment)Universal Fit, Qiagen
FormatRack
Product LineQiagen
SterilitySterile
Tip StyleConductive
Volume (Metric)1100 μL
Unit SizeCase of 960