95401.H4POOL (yeast deletion pools)
Catalog number: 95401.H4POOL
Invitrogen™  Related applications: Genotyping & Genomic Profiling
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
The Yeast Deletion Pools allow simultaneous analysis of large numbers of deletion strains through selective growth conditions. Identification and quantitation of the deletions surviving selection can be determined by hybridizing the deletions to a microarray containing all the barcode oligos (Figure 1). The barcode oligos are unique to each deletion, allowing rapid, high-throughput identification of the deletions. Each Yeast Deletion Pool contains all the deletions within the strain, equally represented within the pool. The set of Yeast Deletion Pools is supplied in five tubes, each containing 200 µl of the pooled deletions.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Shipping Condition: Dry Ice
Contents & storage
Store in ultra-cold freezer (-68 to -85°C).
Documents
Manuals & protocols