Hydrochloric acid, 36.5-38.0%, NF, Multi-Compendial, GMP, J.T.Baker™
Avantor™

Hydrochloric acid, 36.5-38.0%, NF, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets JP, BP, USP, and EP Requirements.
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
47 lb.
140 lb.
475 lb.
Packaging:
View all
Poly pail (Jerrican)
Catalog number 9544-07
Price (EUR)
-
Quantity:
47 lb.
Packaging:
Poly pail (Jerrican)
This product is GMP Manufactured and meets JP, BP, USP, and EP Requirements.
This product is GMP Manufactured and meets JP, BP, USP, and EP Requirements.
Specifications
Boiling Point110°C (1013 hPa)
CAS7647-01-0
ColorColorless
Melting Point-30°C
Molecular FormulaHCl
PackagingPoly pail (Jerrican)
SynonymMuriatic Acid, Hydrogen Chloride, aqueous
GradeMulti-Compendial/NF
Molecular Weight (g/mol)36.46
Percent Purity36.5-38.0%
Quantity47 lb.
Unit SizeEach