Hydrochloric acid, 36.5-38.0%, NF, Multi-Compendial, GMP, J.T.Baker™
Hydrochloric acid, 36.5-38.0%, NF, Multi-Compendial, GMP, J.T.Baker™
Avantor™

Hydrochloric acid, 36.5-38.0%, NF, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets JP, BP, USP, and EP Requirements.
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
47 lb.
140 lb.
475 lb.
Packaging:
View all
Clear glass bottle, PVC-coated
Clear glass bottle
Clear glass bottle, single shipper
Catalog number 9544-33
Price (USD)
-
Request A Quote
Quantity:
2.5 L
Packaging:
Clear glass bottle, PVC-coated
This product is GMP Manufactured and meets JP, BP, USP, and EP Requirements.
This product is GMP Manufactured and meets JP, BP, USP, and EP Requirements.
Specifications
Boiling Point110°C (1013 hPa)
CAS7647-01-0
ColorColorless
Melting Point-30°C
Molecular FormulaHCl
PackagingClear glass bottle, PVC-coated
SynonymMuriatic Acid, Hydrogen Chloride, aqueous
GradeMulti-Compendial/NF
Molecular Weight (g/mol)36.46
Percent Purity36.5-38.0%
Quantity2.5 L
Unit SizeEach