The Applied Biosystems TaqMan SARS-CoV-2 Plus Control is a synthetic RNA positive control that contains target sequences for the SARS-CoV-2 ORF1a, ORF1b, and N gene RUO assays included in the: • TaqMan SARS-CoV-2 with RNase P Assay 2.0 (Cat. No. A51331) • TaqMan SARS-CoV2 MS2 RT-PCR Assay 2.0 (Cat. No. A51327) • TaqMan SARS-CoV2 Fast PCR Assay 2.0 (Cat. No. A51246)
The control should be diluted in TaqMan SARS-CoV-2 Control Dilution Buffer (Cat. No. A49889).
Specifications
Format
Tube
For Use With (Application)
TaqManTM SARS-CoV-2 with Rse P Assay 2.0 TaqManTM SARS-CoV-2 MS2 RT-PCR Assay 2.0 TaqMan™ SARS-CoV2 Fast PCR Assay 2.0