Histostain-SP IHC Kit, rabbit

Catalog number: 956143B

Novex™  Related applications: Anatomical Pathology

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Histostain-SP (Streptavidin-Peroxidase) and Histostain-SAP (Streptavidin-Alkaline Phosphatase) immunohistochemical staining kits employ the LAB-SA (Labeled [strept]Avidin-Biotin) method. For Histostain-SP Kits, DAB or AEC chromogen/substrates generate a colored precipitate around the target antigen in human samples. DAB (brown) is approximately twice as sensitive as AEC (reddish). For Histostain-SAP Kits, AP-Red or BCIP/NBT chromogen/substrates generate a colored precipitate around the target antigen in human samples. BCIP/NBT (dark blue/purple) is about twice as sensitive as AP-Red (red/magenta).

Reactivity: Rabbit

Applications: Immunohistochemistry Detection
For In Vitro Diagnostic Use.

Specifications

Detection Method: Colorimetric
For Use With (Equipment): Light Microscope
Product Line: HISTOSTAIN®
Product Size: 1,000 slides
Reactivity: Rabbit
Substrate: AEC
Substrate Type: HRP (Horseradish Peroxidase) Substrate
Target Species: Human
Target Specificity: Not Target-Specific
Technique: LAB-SA (Labeled Streptavidin-Biotin)
Shipping Condition: Wet Ice

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols