AcroMetrix™ MultiMix FFPE Control (RUO)
Thermo Scientific™  Related applications: Diagnostic Testing
Assess the performance of methods for the isolation, purification, and detection of somatic genetic variants in research oncology testing with the Thermo Scientific™ AcroMetrix™ MultiMix FFPE Controls (RUO). These controls consist of five single-use samples of a synthetic tissue that has been formalin-fixed, paraffin-embedded and sectioned to 10 microns thick. The controls are for Research Use Only and are not for use in diagnostic procedures.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
Showing {{showMoreLimit}} of {{commerceData.products.length}} |  Showing {{shownProducts.length}} of {{commerceData.products.length}} | 
Back to top
Description
Assess the performance of methods for the isolation, purification, and detection of somatic genetic variants in research oncology testing with the Thermo Scientific™ AcroMetrix™ MultiMix FFPE Controls (RUO). These controls consist of five single-use samples of a synthetic tissue that has been formalin-fixed, paraffin-embedded and sectioned to 10 microns thick. The controls are for Research Use Only and are not for use in diagnostic procedures.
For Research Use Only. Not for use in diagnostic procedures.
Documents
Manuals & protocols