Nunc™ Microscope Slides, polystyrene - FAQs
Nunc™ Microscope Slides, polystyrene - FAQs
View additional product information for Nunc™ Microscope Slides - FAQs (960004, 160005)
1 product FAQs found
Which fixation reagents can I use with Permanox plastic?
We have a Technical Bulletin (link below) describing fixation reagents that are compatible with Permanox plastic: https://assets.thermofisher.com/TFS-Assets/LSG/Application-Notes/D19581.pdf