AcroMetrix™ MSSA Positive Control
Thermo Scientific™  Related applications: Diagnostic Testing
Assess the performance of quantitative and qualitative molecular test procedures for the determination of MSSA DNA with the Thermo Scientific™ AcroMetrix™ MSSA Positive Control. Routine use of external run controls enables laboratories to monitor day-to-day test variation, lot-to-lot performance of test kits, and operator variation and can assist in identifying increases in random or systematic error. The control is for In Vitro Diagnostic Use.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
Showing {{showMoreLimit}} of {{commerceData.products.length}} |  Showing {{shownProducts.length}} of {{commerceData.products.length}} | 
Back to top
Description
Assess the performance of quantitative and qualitative molecular test procedures for the determination of MSSA DNA with the Thermo Scientific™ AcroMetrix™ MSSA Positive Control. Routine use of external run controls enables laboratories to monitor day-to-day test variation, lot-to-lot performance of test kits, and operator variation and can assist in identifying increases in random or systematic error. The control is for In Vitro Diagnostic Use.
For In Vitro Diagnostic Use.
Documents
Manuals & protocols