Thermo Scientific™

AcroMetrix™ HCV Controls (RUO)

Catalog number: 960203
Thermo Scientific™

AcroMetrix™ HCV Controls (RUO)

Catalog number: 960203
Monitor day-to-day test variation, lot-to-lot performance of test kits, and operator variation with the Thermo Scientific™ AcroMetrix™ HCV Controls (RUO). These controls identify random or systematic errors in nucleic acid test procedures for the quantitative and qualitative determination of hepatitis C viral RNA (HCV RNA) and can assist in identifying increases in random or systematic error. The AcroMetrix HCV Controls (RUO) are not intended for use with assays used to screen the blood supply. The controls are for Research Use Only and are not for use in diagnostic procedures.
 
Catalog Number
960203
Unit Size
-
Level
High
Packaging
5 x 1.2mL
Price (USD)
Full specifications
DescriptionAcroMetrix HCV High Control
Detectable AnalytesHCV
TypeHCV Control
LevelHigh
Packaging5 x 1.2mL
Quantity5 x 1.2 mL
Unit Size-
Catalog Number
Unit Size
Each
Level
Low
Packaging
5 x 1mL
Price (USD)
Full specifications
DescriptionAcroMetrix NAC HCV Low Control
Detectable AnalytesHCV
TypeHCV Control
LevelLow
Packaging5 x 1mL
Quantity5 x 1 mL
Unit SizeEach
Catalog Number
Unit Size
Each
Level
Mid
Packaging
5 x 1mL
Price (USD)
Full specifications
DescriptionAcroMetrix NAC HCV Mid Control
Detectable AnalytesHCV
TypeHCV Control
LevelMid
Packaging5 x 1mL
Quantity5 x 1 mL
Unit SizeEach
Showing 3 of 3
Catalog NumberSpecificationsUnit SizeLevelPackagingPrice (USD)
960203Full specifications
-High5 x 1.2mLRequest A Quote
DescriptionAcroMetrix HCV High Control
Detectable AnalytesHCV
TypeHCV Control
LevelHigh
Packaging5 x 1.2mL
Quantity5 x 1.2 mL
Unit Size-
950201Full specifications
EachLow5 x 1mLRequest A Quote
DescriptionAcroMetrix NAC HCV Low Control
Detectable AnalytesHCV
TypeHCV Control
LevelLow
Packaging5 x 1mL
Quantity5 x 1 mL
Unit SizeEach
950202Full specifications
EachMid5 x 1mLRequest A Quote
DescriptionAcroMetrix NAC HCV Mid Control
Detectable AnalytesHCV
TypeHCV Control
LevelMid
Packaging5 x 1mL
Quantity5 x 1 mL
Unit SizeEach
Showing 3 of 3
Monitor day-to-day test variation, lot-to-lot performance of test kits, and operator variation with the Thermo Scientific™ AcroMetrix™ HCV Controls (RUO). These controls identify random or systematic errors in nucleic acid test procedures for the quantitative and qualitative determination of hepatitis C viral RNA (HCV RNA) and can assist in identifying increases in random or systematic error. The AcroMetrix HCV Controls (RUO) are not intended for use with assays used to screen the blood supply. The controls are for Research Use Only and are not for use in diagnostic procedures.
For Research Use Only. Not for use in diagnostic procedures.