This product will be integrating with a quality controls portfolio that includes Thermo Scientific™ AcroMetrix™ molecular controls, MAS™ clinical chemistry controls, and other products for your clinical testing workflow. Learn more about how this transition may affect your QC products
QConnect HBVDNA+ Control
Catalog number: 960243
Applied Biosystems™  Related applications: Clinical & Translational Research | Diagnostic Testing
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
QConnect HBVDNA+ Control (formerly AcroMetrix® PeliSpy™ NAT HBV Low Control) is a low-level, external quality control designed to help laboratories assess and monitor the performance of assays that detect hepatitis B virus (HBV) DNA. This control has been optimized for use with newer assay systems with higher sensitivities.
Manufactured in accordance with ISO 17511, this control is designed to yield consistent results from lot to lot and is ideal for monitoring assay performance over long periods of time. Routine use enables laboratories to monitor day-to-day test variation, lot-to-lot performance of test kits, and operator variation.
For In Vitro Diagnostic Use.
Test Type: Serology Testing, Infectious Diseases
Control Type: Serology Control, Positive Control, Molecular Control
Product Size: 50 vials
Target Organism Class: Hepatitis B Virus (HBV)
Contents & storage
Store unopened vials at 20°C or lower. May be stored at 2-8°C for up to 5 days, or at room temperature for up to 24 hours prior to use.
Manuals & protocols