Orion™ Ionplus™ Sure-Flow™ Cyanide Combination Ion Selective Electrode
Orion™ Ionplus™ Sure-Flow™ Cyanide Combination Ion Selective Electrode
Orion™ Ionplus™ Sure-Flow™ Cyanide Combination Ion Selective Electrode
Orion™ Ionplus™ Sure-Flow™ Cyanide Combination Ion Selective Electrode
Orion™ Ionplus™ Sure-Flow™ Cyanide Combination Ion Selective Electrode
Thermo Scientific™
Orion™ Ionplus™ Sure-Flow™ Cyanide Combination Ion Selective Electrode
Measure cyanide ions in aqueous solutions quickly, simply, accurately and economically.
Have Questions?
Catalog NumberConnector Types
9606BNWPBNC (Waterproof)
Catalog number 9606BNWP
Price (MXN)
-
Connector Types:
BNC (Waterproof)
Measure cyanide ions in aqueous solutions quickly, simply and economically with Thermo Scientific™ Orion™ Cyanide Electrodes.
The ThermoFisher Orion Cyanide electrode complies with SM 4500-CN- F-1999 for Drinking water ; and SM 4500-CN- F-1999, ASTM D2036-09 (A)Wastewater measurements.
Features
  • 9606BNWP is a combination electrode with a Sure-Flow™ junction. When conventional electrode junctions clog, the Sure-Flow junction can be opened and flushed clean simply by pressing the cap.
Solutions required
  • ISA
    • 951011 ISA for cyanide measurements, 475 mL bottle
  • Electrode fill solution
    • 900062 Optimum Results B electrode filling solution, 5 x 60 mL bottles
Items included with
  • 900062 Optimum Results B electrode filling solution. 60mL 1 bottle
  • 948201 Polishing strip. Qty 2
Warranty period
  • 12 months from date of purchase
Specifications
DescriptionModel 9606BNWP; Cyanide combination
Connector TypesBNC (Waterproof)
For Use With (Equipment)ISE Meter
JunctionSure-Flow Junction
MaterialEpoxy
Temperature (Metric) Operating0°C to 80°C
TypeIon Selective Electrode
Unit SizeEach