Olink™ Reveal Kit - FAQs
Olink™ Reveal Kit - FAQs
View additional product information for Olink™ Reveal Kit - FAQs (96200)
13 product FAQs found
What do I need to get started with Olink Reveal?
To get started with Olink Reveal, you’ll first need to purchase the kit. Once your order is placed, a Field Application Scientist will reach out to you provide everything you need, including access to NPX Map software, information on optional virtual training, and the required ngs2counts software and custom recipe for your next-generation sequencing system. All the guidance and resources to run Olink Reveal will be delivered directly following your purchase.
Where can I find more information about Olink Reveal?
For detailed information, videos, and more, visit this page.
Where can I find the Safety Data Sheet (SDS) for Olink Reveal?
SDS documents for Olink Reveal can be accessed here.
Is there validation data available for Olink Reveal?
Yes. Olink Reveal is based on widely-used PEA technology and has undergone rigorous analytical and technical validation to ensure reproducibility, specificity, and performance across studies. You can find an overview of the validation process here and detailed data for each individual protein here.
What proteins are included in the Olink Reveal panel?
For a full list of the 1034 proteins, download this document.
Who will support me if I have questions or need help?
During our integration, you can connect with either techsupport@thermofisher.com or support@olink.com for guidance. Our support teams will collaborate to ensure you receive the assistance you need.
How do I request a quote for Olink Reveal?
Click the ''Request a Quote'' button on the product page and fill out the necessary fields. A representative from Thermo Fisher Scientific will get in touch with you, and a quote will be tailored to your project needs and sample volume.
How do I get in touch to learn more about Olink Reveal?
Click the ''Request a Quote'' button on the product page and fill out the necessary fields. A representative from Thermo Fisher Scientific will get in touch with you to discuss your needs.
Is there a difference between buying Olink Reveal from Olink vs. Thermo Fisher Scientific?
No. The product and pricing are the same whether you purchase Olink Reveal through Olink or Thermo Fisher Scientific.
What other products does Olink offer? Can I buy them through Thermo Fisher Scientific?
Olink offers a range of panels in different sizes, with options for absolute quantification. You can compare Olink panels by visiting: https://olink.com/products/compare
Currently, only Olink Reveal can be purchased through Thermo Fisher Scientific.
Can I purchase Olink Reveal through Thermo Fisher Scientific?
Olink is now part of Thermo Fisher Scientific. This integration brings Olink’s proven and transformative proteomics solutions into Thermo Fisher Scientific’s leading life sciences portfolio, making it easier for more scientists to access Olink Reveal and accelerate discovery and scientific breakthroughs.
Who is Olink? What is PEA technology?
Olink, now part of Thermo Fisher Scientific, is a global leader in proteomics, with a mission to accelerate protein research together with the scientific community to understand real-time biology and gain actionable insights into human health and disease. The Olink Proximity Extension Assay (PEA) technology uniquely combines specificity, sensitivity, and scalability for high-throughput, multiplex protein biomarker analysis. Learn more here.