Sulfuric acid, 95.0-98.0% (by acidimetry), NF, Multi-Compendial, GMP, J.T.Baker™
Avantor™

Sulfuric acid, 95.0-98.0% (by acidimetry), NF, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets N.F., EP, and BP Requirements.
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
500 mL
2.5 L
Número de catálogo 9671-02
Precio (EUR)
-
Cantidad:
500 mL
This product is GMP Manufactured and meets N.F., EP, and BP Requirements.
Especificaciones
Punto de ebullición∼335°C (1013 hPa)
CAS7664-93-9
Nombre del producto químico o materialSulfuric acid
ColorColorless
Punto de fusión3°C
Fórmula molecularH2SO4
EnvaseClear glass bottle, single shipper
SinónimoSulfuric acid
GradoMulti-Compendial/NF
Peso molecular (g/mol)98.08
Porcentaje de pureza95.0-98.0%
Cantidad500 mL
Unit SizeEach