Thermo Scientific™

AcroMetrix Serology Single Analyte Controls

Catalog number: 969035
Thermo Scientific™

AcroMetrix Serology Single Analyte Controls

Catalog number: 969035
Estimate the precision and reproducibility of serological assays that test human plasma from donations of whole blood and blood components with the Thermo Scientific™ AcroMetrix Serology Single Analyte Controls. These controls mimic naturally occurring plasma specimens with low reactivity of a single analyte and enable monitoring of day-to-day test performance of IVD assays for the detection of human infectious diseases. The controls are for In Vitro Diagnostic Use.
 
Catalog Number
969035
Unit Size
Each
Description
AcroMetrix Syphilis Control
Packaging
5 x 1mL
Price (USD)
Full specifications
DescriptionAcroMetrix Syphilis Control
Detectable AnalytesSyphilis
Sample TypePlasma
TypeControl
Packaging5 x 1mL
Unit SizeEach
Catalog Number
Unit Size
Each
Description
AcroMetrix Trisceen Control
Packaging
10 x 1mL
Price (USD)
Full specifications
DescriptionAcroMetrix Trisceen Control
Detectable AnalytesHBV, HIV, HCV
Sample TypePlasma
TypeControl
Packaging10 x 1mL
Quantity10 x 1 mL
Unit SizeEach
Catalog Number
Unit Size
Each
Description
AcroMetrix HIVP24 Control
Packaging
5 x 5mL
Price (USD)
Full specifications
DescriptionAcroMetrix HIVP24 Control
Detectable AnalytesHIV
Sample TypePlasma
TypeControl
Packaging5 x 5mL
Quantity5 x 5 mL
Unit SizeEach
Catalog Number
Unit Size
Each
Description
AcroMetrix Anti-HBs Control
Packaging
5 x 1mL
Price (USD)
Full specifications
DescriptionAcroMetrix Anti-HBs Control
Detectable Analytesanti-HBs
Sample TypePlasma
TypeControl
Packaging5 x 1mL
Quantity5 x 1 mL
Unit SizeEach
Showing 4 of 4
Catalog NumberSpecificationsUnit SizeDescriptionPackagingPrice (USD)
969035Full specifications
EachAcroMetrix Syphilis Control5 x 1mLRequest A Quote
DescriptionAcroMetrix Syphilis Control
Detectable AnalytesSyphilis
Sample TypePlasma
TypeControl
Packaging5 x 1mL
Unit SizeEach
960811Full specifications
EachAcroMetrix Trisceen Control10 x 1mLRequest A Quote
DescriptionAcroMetrix Trisceen Control
Detectable AnalytesHBV, HIV, HCV
Sample TypePlasma
TypeControl
Packaging10 x 1mL
Quantity10 x 1 mL
Unit SizeEach
960038Full specifications
EachAcroMetrix HIVP24 Control5 x 5mLRequest A Quote
DescriptionAcroMetrix HIVP24 Control
Detectable AnalytesHIV
Sample TypePlasma
TypeControl
Packaging5 x 5mL
Quantity5 x 5 mL
Unit SizeEach
960855Full specifications
EachAcroMetrix Anti-HBs Control5 x 1mLRequest A Quote
DescriptionAcroMetrix Anti-HBs Control
Detectable Analytesanti-HBs
Sample TypePlasma
TypeControl
Packaging5 x 1mL
Quantity5 x 1 mL
Unit SizeEach
Showing 4 of 4
Estimate the precision and reproducibility of serological assays that test human plasma from donations of whole blood and blood components with the Thermo Scientific™ AcroMetrix Serology Single Analyte Controls. These controls mimic naturally occurring plasma specimens with low reactivity of a single analyte and enable monitoring of day-to-day test performance of IVD assays for the detection of human infectious diseases. The controls are for In Vitro Diagnostic Use.
For In Vitro Diagnostic Use.