QConnect NAT Blood Screening Controls
Thermo Scientific™  Related applications: Diagnostic Testing
Estimate the precision and reproducibility of NAT assays that are designed to detect viral nucleic acids in human plasma from donations of whole blood and blood components for transfusion with the Thermo Scientific™ QConnect NAT Blood Screening Controls. These controls mimic naturally occurring plasma specimens and can help detect potential systematic deviations of tests for the qualitative determination of viral nucleic acids. The controls are for In Vitro Diagnostic Use.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
Showing {{showMoreLimit}} of {{commerceData.products.length}} |  Showing {{shownProducts.length}} of {{commerceData.products.length}} | 
Back to top
Description
Estimate the precision and reproducibility of NAT assays that are designed to detect viral nucleic acids in human plasma from donations of whole blood and blood components for transfusion with the Thermo Scientific™ QConnect NAT Blood Screening Controls. These controls mimic naturally occurring plasma specimens and can help detect potential systematic deviations of tests for the qualitative determination of viral nucleic acids. The controls are for In Vitro Diagnostic Use.
For In Vitro Diagnostic Use.
Documents
Manuals & protocols