KingFisher™ Plastics for 96 standard and PCR formats
Thermo Scientific™
KingFisher™ Plastics for 96 standard and PCR formats
Optimize your performance using Thermo Scientific™ KingFisher consumables designed specifically for use with the KingFisher Flex, Duo Prime, and Presto instruments.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Quantity
97002540BKingFisher Apex48/Cs.
97002560KingFisher Apex50/Cs.
97002540KingFisher Flex and Presto48/Cs.
97002524KingFisher Flex100/Cs.
97002514KingFisher Flex80/Cs.
97002524BKingFisher Apex100/Cs.
Catalog number 97002540B
Price (USD)
225.65
Online exclusive
269.00
Save 43.35
Case of 48
For Use With (Equipment):
KingFisher Apex
Quantity:
48/Cs.
Price (USD)
225.65
Online exclusive
269.00
Save 43.35
Case of 48
Optimize your performance using Thermo Scientific™ KingFisher Plastics designed specifically for use with the KingFisher Flex, Duo Prime, Presto and Apex instruments. KingFisher instruments utilize disposable plastic plates and tip combs made of polypropylene, which are ideal for magnetic particle processing due to their low binding affinity for biomolecules. Both the yield and quality of the isolated protein or nucleic acid are significantly improved with special plates and tip combs designed for KingFisher systems.
Use With: Thermo Scientific™ KingFisher instruments, including KingFisher Apex, KingFisher Flex and KingFisher Presto.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
For Use With (Equipment)KingFisher Apex
Quantity48/Cs.
Capacity200 μL
Unit SizeCase of 48
Contents & Storage
KingFisher 96 Plate (200 μL), Barcoded