Search
Search

Use for predictable HPLC separations of compounds in highly aqueous mobile phases.
| Número do catálogo | Length (Metric) | Diameter (Metric) | Particle Size |
|---|---|---|---|
| 97303-103030 | 100 mm | 3 mm | 3 μm |
| 97305-102130 | 100 mm | 2.1 mm | 5 μm |
| 97305-104630 | 100 mm | 4.6 mm | 5 μm |
| 97305-013001 | 10 mm | 3 mm | 5 μm |
| 97305-012101 | 10 mm | 2.1 mm | 5 μm |
| 97303-154630 | 150 mm | 4.6 mm | 3 μm |
| 97303-104630 | 100 mm | 4.6 mm | 3 μm |
Ensure consistent, predictable liquid chromatography (LC) separations of polar analytes using Thermo Scientific™ Syncronis™ aQ Columns. High surface area silica and dense stationary phase coverage minimize secondary interactions and deliver reproducible high-resolution separations. The polar end-capped C18 phase provides optimal retention of polar analytes and is fully compatible with 100% aqueous mobile phases. Manufactured and rigorously tested in ISO 9000 facilities, these columns are designed to meet a range of analytical needs including fast method development, quantitative analysis, and quality control.