Ammonia solution, 27.0-30.0%, NF, Multi-Compendial, GMP Manufactured Product, J.T.Baker™
Avantor™

Ammonia solution, 27.0-30.0%, NF, Multi-Compendial, GMP Manufactured Product, J.T.Baker™

This product is GMP Manufactured and meets N.F., EP, and BP Requirements.
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
2.5 L
360 lb.
Número de catálogo 9736-05
Precio (EUR)
-
Cantidad:
2.5 L
This product is GMP Manufactured and meets N.F., EP, and BP Requirements.
Especificaciones
Punto de ebullición-33°C (1013 hPa)
CAS1336-21-6
Nombre del producto químico o materialAmmonia solution
ColorColorless
Punto de fusión-77°C
Fórmula molecularNH3
EnvaseClear glass bottle, single shipper
SinónimoAmmonium hydroxide
GradoMulti-Compendial/NF
Peso molecular (g/mol)35.05
Porcentaje de pureza27.0-30.0%
Cantidad2.5 L
Unit SizeEach