Thermo Scientific™
Richard-Allan Scientific™ Fast Flex™ Holding Solution
Related applications:
Anatomical Pathology
Employ a fixative designed for use in the grossing room with the Thermo Scientific™ Richard-Allan Scientific™ Fast Flex™ Holding Solution.
Back to top
  • Place cassettes in holding solution for at least 20 minutes prior to loading on the processor
  • Compatible with 10% Neutral Buffered Formalin and other fixatives