Search
Search

| Catalog Number | Description |
|---|---|
| 981695 | HbA1c Control Normal; Kit size: 5 x 1mL |
| 980997 | SpeciCal Protein Calibrator; Kit size: 5 x 1mL |
| 981250 | Specitrol; Kit size: 5 x 1mL |
| 981252 | RF Control; Kit size: 5 x 1mL |
| 981659 | Hemolyzing Reagent; Kit size: 1000mL |
| 981696 | HbA1c Control Abnormal; Kit size: 5 x 1mL |
| 981799 | Specitrol High; Kit size: 5 x 1mL |
| 981915 | Lipoprotein(a); Kit Size: 1 x 1mL antiserum |
| 981920 | Rheumatoid factors 2 (RF 2); Kit Size: 2 x 6mL antiserum |
| 981924 | HbA1c Pretreatment liquid; Kit size: 12 x 20mL |
| 981936 | IgA; Kit Size: 2 x 3mL antiserum |
| 981937 | IgG; Kit Size: 2 x 3mL antiserum |
| 981938 | IgM; Kit Size: 2 x 3mL antiserum |
| 981943 | Transferrin; Kit Size: 2 x 3mL antiserum |
| 981917 | Lipoprotein(a) Control; Kit size: 3 x 1mL |
| 981946 | D-Dimer, Kit Size: 2 x 8mL |
| 981945 | sTfR; Kit Size: 2 x 5mL |
| 981871 | D-Dimer Calibration Set |
| 981868 | D-Dimer Control |
| 981869 | D-Dimer Control High |
| 981874 | Ferritin Calibration Set |
| 981875 | Ferritin Control |
| 981876 | Ferritin Control High |
| 981865 | sTfR Calibration Set |
| 981866 | sTfR Control |
| 981867 | sTfR Control High |
| 981948 | HbA1c; Kit Size: 2 x 17.1mL total for HbA1c |
| 981947 | HbA1c; Kit Size: 6 x 17, 1mL total for HbA1c |
| 981863 | HbA1c Calibrator |
Receive best-in-class methodologies and standardized performance with Thermo Scientific™ Indiko™ and Konelab™ System Specific Protein Reagents, which include a specific protein test menu of 15 immunoturbidimetric methods. All reagents are ready to go and packaged in volume-optimized, barcoded kits; in-house evaluation as well as optimization of reagents guarantees continuous support and optimum results.
All Indiko and Konelab methods are based on carefully-designed protocols for barcoded system reagents. Calibrators and controls are available in separate kits.
Highlights:
Ordering Information: Not available in the U.S.A.
Specific Protein Applications for Indiko and Konelab Analyzers: