Indiko™ and Konelab™ System Clinical Chemistry Reagents, GGT to Uric Acid
Actual product may vary
Thermo Scientific™

Indiko™ and Konelab™ System Clinical Chemistry Reagents, GGT to Uric Acid

Fully standardize performance with Thermo Scientific™ Indiko™ and Konelab™ System Clinical Chemistry Reagents.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDetectable AnalytesQuantity
981818Urea10 x 60 mL
981236Iron10 x 20 mL
981301Triglycerides6 x 60 mL
981377Gamma-GT10 x 20 mL
981391Uric Acid7 x 60 mL
981778Gamma-GT15 x 15 mL
981786Triglycerides12 x 20 mL
981788Uric Acid8 x 20 mL
981827Total Protein Plus12 x 20 mL
981843U/CSF Protein8 x 20 mL
981846Lipase4 x 25 mL
981890Phosphorus6 x 18 mL
981891Phosphorus8 x 10 mL
981905Magnesium8 x 6 mL
Catalog number 981818
Price (MXN)
-
Detectable Analytes:
Urea
Quantity:
10 x 60 mL
Enhance your laboratory's performance with Thermo Scientific™ Indiko™ and Konelab™ System Clinical Chemistry Reagents. These ready-to-use assays come fully validated and compliant, featuring automatic barcoding for efficient process and reordering checks. System reagents provide timely and accurate information, helping to ensure results you can stand behind.
  • Indiko™ and Konelab™ applications utilize carefully selected methods according to IFCC and other widely used recommendations
  • Clinical chemistry methods are provided with system specified multi-calibrators and -controls with traceability to international reference materials
  • The CE marked products are compliant with EU regulatory requirements for in vitro diagnostic medical devices
Ordering Information: Not available in the U.S.A.
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Certifications/ComplianceCE Marked
DescriptionUrea; Kit Size: 10 x 60mL
Quantity10 x 60 mL
Sample TypeSerum
TypeReagent
CE MarkerYes
Detectable AnalytesUrea
FormLiquid
Unit SizeEach