Search
Search

| Catalog Number | Description |
|---|---|
| 981927 | Albumin MST; Kit Size: 2 x 3mL antiserum |
| 981251 | CRP Control; Kit size: 5 x 1mL |
| 981674 | CRP Calibration Set; Kit size: 5 x 1mL |
| 981679 | CRP Control High; Kit size: 5 x 1mL |
| 981699 | CRP; Kit Size: 2 x 3mL antiserum |
| 981798 | CRP High Sensitivity; Kit Size: 1 x 4mL antiserum |
| 981852 | CRP High Sensitivity Control; Kit size: 3 x 1mL |
| 981877 | Albumin U Calibrator; Kit Size: 2 x 2mL |
| 981878 | Albumin U Control; Kit size: 2 x 2mL |
| 981879 | Albumin U Control High; Kit size: 2 x 2mL |
| 981921 | APO A1/APO B Calibrator; Kit size: 2 x 1mL |
| 981929 | Apolipoprotein A1; Kit Size: 2 x 3mL antiserum |
| 981930 | Apolipoprotein B; Kit Size: 2 x 3mL antiserum |
| 981933 | CRP; Kit Size: 1 x 3mL antiserum |
| 981932 | Complement C4; Kit Size: 2 x 3mL antiserum |
| 981931 | Complement C3; Kit Size: 2 x 3mL antiserum |
All Indiko and Konelab methods are based on carefully-designed protocols for barcoded system reagents. Calibrators and controls are available in separate kits.
Highlights:
Ordering Information: Not available in the U.S.A.
Specific Protein Applications for Indiko and Konelab Analyzers: