Gallery™ System Reagents for Environmental and Industrial Analysis
Gallery™ System Reagents for Environmental and Industrial Analysis
Gallery™ System Reagents for Environmental and Industrial Analysis
Thermo Scientific™
Gallery™ System Reagents for Environmental and Industrial Analysis
Select from these ready-to-use reagents and enzymatic kits specifically designed for environmental and industrial water analysis using a Thermo Scientific™ Gallery™ Discrete Analyzer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeNo. of Tests
984707Iron (Ferrous) System Reagent1080
984187TON Enzymatic Reduction System Reagent275
984350TON-V Vanadium Reduction System Reagent360
984351TON-V Vanadium Reduction System Reagent500
984357Chromium System Reagent480
984358Magnesium System Reagent350
984362Ammonia System Reagent2000
984363Ammonia System Reagent1300
984364Chloride System Reagent500
984365Chloride System Reagent2500
984366Phosphate System Reagent2300
984367Phosphate System Reagent11700
984368Phosphate System Reagent3000
984369TON System Reagent1300
984370TON System Reagent750
984371TON System Reagent1000
984372TON System Reagent5000
984620Total Hardness System Reagent1100
984621Total Hardness System Reagent975
984622Total Hardness System Reagent4300
984623Alkalinity System Reagent975
984624Alkalinity System Reagent780
984625Silica System Reagent970
984626Silica System Reagent R21950
984627Silica System Reagent R31950
984629Fluoride System Reagent R12200
984630Fluoride System Reagent4800
984631Fluoride System Reagent R32200
984648Sulphate System Reagent1100
984649Sulphate System Reagent5100
984652TON System Reagent3400
984706Iron (Ferrous) System Reagent1900
Catalog number 984707
Price (MXN)
-
Type:
Iron (Ferrous) System Reagent
No. of Tests:
1080
Save time and reduce errors by eliminating reagent preparation with the Thermo Scientific™ Gallery™ system reagents for environmental testing (clean water, wastewater, soil and sludge digests, effluents, and saline) and industrial (process water) analysis. With over 40 ready-to-use Gallery system reagents, these kits include 1 to 3 reagents in 20 mL barcoded vials, allowing the necessary identification data, such as lot and expiration date, to be read automatically by the Gallery Discrete Analyzer's barcode reader. Automated dilutions make it possible to apply methods for a wide range of analyte concentrations and the test applications are based on recommended standard water and wastewater methods used in most countries.
Highlights:
  • All test applications are based on recommended standard methods in water and wastewater analysis of different countries
  • Kit structure includes 1 to 3 reagents in 20 mL vials
  • Barcoded vials are simple and fast to use
  • All necessary data, such as lot and expiration date, are automatically read
  • Automated dilutions make it possible to apply methods for a wide range of analyte concentrations
  • Types of matrices analyzed include clean water, wastewater, soil and sludge digests, effluents, saline and process water
Specifications
Capacity (Metric)20 mL
DetectsIron (Ferrous)
For Use With (Equipment)Gallery Discrete Analyzer
Interference Filter510/880
No. of Tests1080
Quantity4 x 20 mL
TypeIron (Ferrous) System Reagent
Unit SizeEach