Gallery™ Discrete Analyzer
Actual product may vary
Gallery™ Discrete Analyzer
Gallery™ Discrete Analyzer
Gallery™ Discrete Analyzer
Gallery™ Discrete Analyzer
Gallery™ Discrete Analyzer
Gallery™ Discrete Analyzer
Gallery™ Discrete Analyzer
Gallery™ Discrete Analyzer
Gallery™ Discrete Analyzer
Gallery™ Discrete Analyzer
Gallery™ Discrete Analyzer
Thermo Scientific™

Gallery™ Discrete Analyzer

Bring excellent analytical performance to colorimetric, enzymatic and electrochemical measurements with the Thermo Scientific™ Gallery™ Automated Photometric Analyzer.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
98610001Gallery Discrete Analyzer without ECM Unit
98611001Gallery Discrete Analyzer with ECM Unit
Catalog number 98610001
Price (MXN)
-
Description:
Gallery Discrete Analyzer without ECM Unit

Designed to increase efficiency for photometric (colorimetric and enzymatic) and electrochemical (pH and conductivity) analysis, the Thermo Scientific™ Gallery™ Discrete Analyzer provides fast, reproducible results in a compact, benchtop design. Its discrete cell technology allows laboratories to measure multiple analytes simultaneously while reducing total analysis and operator time. All necessary analysis steps are automated and low detection levels can be achieved. The analyzer includes a combined sample and reagent disk for a maximum capacity of 90 samples and 30 reagents, with the ability to perform as many as 200 tests/hour. Achieve increased analytical performance in a wide range of applications including food and beverage analysis, environmental testing, and industrial quality control.

Automated operation and reliable technology

  • Continuous access to samples, reagents, and cuvettes without interruption of routine workflows.
  • Integrated platform for photometric and electrochemical analysis can be operated simultaneously.
  • All necessary analysis steps are automated, enabling up to 2 hours walk-away time for the operator.
  • Increased throughput with a sample capacity of up to 200 tests/hour.
  • Dilutions and repeat analyses are handled automatically.
  • Minimal sample pretreatment required.

Robust capability 

  • Large concentration range measurement, with excellent reproducibility.
  • User-specific applications can be developed to meet individual testing needs.
  • Combined sample and reagent disk with up to 90 sample positions on-board.
  • Several blanking possibilities to eliminate sample matrix effects.
  • Ability to achieve detection of very low levels (ppb).
  • Wavelength range from 340nm to 880nm.
  • Easy dilution management.

Compact design and economical

  • System reagents eliminate time-consuming reagent preparation, freeing operators for other tasks.
  • Unique low volume cuvette design for less reagent waste and potentially lower operating costs.
  • Open system – modify existing method or create your own test.
  • Contamination-free analysis using disposable cuvettes.
  • Optional ECM-unit for conductivity and pH.
  • No need for an external source of water.

Versatile and user-friendly 

  • Accommodates a variety of sample cups and primary tubes.
  • No risk of carry-over or requirement to wash glassware.
  • No additional priming or method changeover time.
  • Identification is automated by a barcode reader.
  • Save space: Self-contained benchtop for small footprint.
  • Automatic startup and shutdown protocols.
  • Intuitive graphic user interface.
Specifications
Analytical MethodColorimetric, Enzymatic and Electrochemical With Automated Photometric Analyzer
CalibrationFactor, Bias, Linear, Logit-Log, Spline, Polynomial and Point-to-Point
CapacityMix of 9 to or 18 to Position Sample Racks
CuvettesDiscrete Disposable Cuvettes. On Board Capacity 360 Measurement Cells.
Depth (English)27.5 in.
Depth (Metric)70 cm
DescriptionGallery Discrete Analyzer without ECM Unit
Distilled Water ConsumptionDeionized Water Consumption 1.5 L/hr.
Frequency50/60 Hz +/-5%
Height (English)24.4/51.1 in. (Cover Closed/Cover Open)
Height (Metric)62/130 cm (Cover Closed/Cover Open)
Measurement MethodPhotometric
Operating SystemAutomated Photometric (Colorimetric and Enzymatic) Analysis
Quality Control DetailsReal-Time QC Program with Multiple and Variable Rules. Control Frequency User-Definable. Out-of-Specification Control Results Flagged.
Reagents6-Position Reagent Racks, Max 6 Racks on-Board
Sample DilutionAutomatic Sample Pre-Dilution. Automatic Sample Dilution of Over-Range Tests with Automatic re-Run. Addition of Manual Pre-Dilution Value for Result Calculation.
Throughput200 Tests/hr.
Voltage100/240 V +/-10%
Weight (Metric)85 kg
Width (English)29.5 in.
Width (Metric)75 cm
TypeDiscrete Analyzer
Unit SizeEach

Frequently asked questions (FAQs)

Can I integrate discrete analysis with ion chromatography for comprehensive water analysis?

Yes. We offer Disc IC packages that pair discrete analysis with Thermo Scientific Dionex ion chromatography systems, delivering high throughput, comprehensive water/ion analysis without method changeover.

How does a discrete analyzer compare to flow analyzers (FIA/SFA/CFA)?

Compared with traditional continuous-flow systems, Gallery and Gallery Plus discrete analyzers’ random-access, discrete design with disposable cuvettes minimizes carryover, cuts reagent use (typically <100 μL per test), shortens startup/changeover times, and boosts throughput (200–350 tests/hr vs 60–120 tests/hr). Labs also report much lower cost-per-test (up to 20x) due to smaller volumes and less waste. These systems also provide more measuring channels (12 filter positions), greater method stability, and fully automated micro liquid handling, which reduce operator skill requirements and system maintenance.

What reagents and consumables are available? Can I use third party reagents?

We offer ready to use, barcoded system reagents and enzymatic kits for environmental/industrial water and food & beverage testing, these minimize prep time, consumption, and cost per analysis. The platform is also open and flexible to support third party photometric methods and reagents if needed.

What “walk away” and automation features are built in?

After loading samples and reagents, the system can run unattended for up to 3 hours. Automation features include the ability to add samples mid-cycle (for the next cycle) without disrupting current tests, automated start up/shut down, and barcode scanning/LIMS connectivity for traceability.

Can I run photometric tests and pH/conductivity at the same time?

Yes. With the optional Electrochemical Measurement (ECM) module, pH (approximately 2 to 12) and conductivity (~20 μS/cm–112 mS/cm) can be measured in parallel with standard photometric assays, enabling truly consolidated multiparameter runs. ECM throughput is up to 54 measurements/hr on Gallery systems and up to 67 measurements/hr on Gallery Plus models.