Gallery Enzyme Master
Gallery Enzyme Master
Gallery Enzyme Master
Gallery Enzyme Master
Thermo Scientific™
Gallery Enzyme Master
Perform robust, and reproducible enzyme assays with automated Gallery Enzyme Master. It efficiently manages many different enzymes and many different measuring conditions—all in a single instrument that offers consistent, reproducible performance.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberElectrochemistry Unit (ECM)
98610004Optional
98611004Included
Catalog number 98610004
Price (MXN)
-
Electrochemistry Unit (ECM):
Optional
Gain confidence in your resultls with the Thermo Scientific™ Gallery™ Enzyme Master - the first automated discrete analyzers designed specifically for enzyme assay applications. Combining robust hardware and new custom-designed software, these best-in-class solutions deliver fully automated incubation settings, reagent additions and precise measurement calculations—all with a touch of a button. They offer the smartest (and fastest) way yet to streamline method development and deliver reliable results from enzyme assay analysis. The optional electrochemistry unit (ECM) allows the parallel determination of pH Optimal up to 67 samples per hour along with other photometric tests.
Confidence in your Results
• Precise temperature control
• Temperature overheating limits
• Accurate incubation time to the seconds
• Superior temperature control and lack of edge effects
Highthroughput Enzyme Assay
• Walkaway solution
• Easy to use, train, and maintain
• Many enzymes, many substrates, many methods—all on a single instrument
• Fully-automated liquid handling (sample, buffer and substrate addition, and mixing)
• Disposable cuvettes (no carryover)
Fully automated workflow
• Easy, automated method development
• Easy method transfer
• Enzyme-specific method parameters all in one central place
Traceability
• Time stamped raw data
• Audit trail
• User administration
Parallel pH Measurement
• ECM Module for parallel pH measurement
• Facilitates method development
• Up to 67 samples pH measurement per hour
Specifications
Analytical MethodDiscrete analysis
Electrochemistry Unit (ECM)Optional
Incubation Temperature25°C to 60°C
Measurement MethodPhotometric: 340nm to 880nm
QuantityEach
Reagent CapacityUp to 30
Sample Size45-90 Samples
Throughput200 Photometric Tests/hr.
Unit SizeEach
Frequently asked questions (FAQs)
What does data integrity and connectivity look like (LIMS/LIS, QC, audit trail) for the Gallery Enzyme Master system?
Gallery Enzyme Master systems provide time stamped raw kinetic data, user administration, and an audit trail, plus CLSI LIS02 A2 interface options. Real time QC supports multiple user definable Westgard rules, and results can be exported to spreadsheets/LIMS for further calculations and reporting.
Why is temperature control important for enzyme analysis, and how does the Gallery Enzyme Master system handle it?
Enzyme activity is temperature sensitive, a 1 °C shift can change measured activity by 4–8%, so precise control matters. The Gallery Enzyme Master discrete analyzer offers smart incubation scheduling (runs from low to high temperature methods to prevent overheating), ±0.3°C control accuracy, to the second incubation timing, and a 25–60 °C incubator range.
How is the Gallery Enzyme Master different from the standard Thermo Scientific Gallery discrete analyzer?
Both are discrete analyzers with similar hardware, but the Gallery Enzyme Master system includes software and workflow features tailored to enzyme assays, such as enzyme specific method parameters (incubation temperature/time, blank strategy, data rate), more stringent temperature management, and a dedicated enzyme filter set. The standard Gallery system is a general purpose photometric/electrochemical platform used broadly across food, beverage, environmental and industrial testing, whereas the Gallery Enzyme Master system is specialized for enzymes and method development/transfer.
What measurement techniques and optics does the Gallery Enzyme Master support?
It supports colorimetric end point and kinetic, turbidimetric, and bichromatic reactions (with or without sample blanking). Optics include a single channel interference filter photometer (12 filter positions) covering 340–880 nm, data collection every nine seconds, and kinetic runs up to one hour. An optional ECM module adds parallel pH and conductivity measurements.
What is the Gallery Enzyme Master and what problem does it solve?
The Gallery Enzyme Master discrete analyzer is a fully automated discrete photometric analyzer purpose‑built for enzyme assay work (activity, assays, and kinetics). It automates reagent additions, incubation time/temperature, and measurements, streamlining method development and routine testing while improving reproducibility versus manual UV‑Vis or microplate workflows.