Gallery™ Plus Beermaster Discrete Analyzer
Actual product may vary
Thermo Scientific™

Gallery™ Plus Beermaster Discrete Analyzer

Save significant time in sample pretreatment and analysis by automating labor-intensive and time-consuming bitterness testing.

Have Questions?
Catalog NumberMeasurement Method
98622001Photometric Analysis, Colorimetric
Catalog number 98622001
Price (BRL)
-
Measurement Method:
Photometric Analysis, Colorimetric

Automate labor-intensive and time-consuming beer, cider, malt, and wort testing with the Thermo Scientific™ Gallery™ Plus Beermaster Discrete Analyzer. Using discrete cell technology, the analyzer simultaneously automates the determination of analytes including beta-glucan, FAN, and SO2 from the same sample. This enables fast turnaround and expansion of your laboratory's analytical capabilities. The Bitterness Column Module (BCM) in the Gallery™ Plus Beermaster Discrete Analyzer enables unique automated pretreatment and measurement for bitterness. The discrete analyzer can accommodate 180 samples and 42 reagents in its separated sample and reagent disks, with the capacity to perform up to 350 tests/hour, and approximately 8 bitterness tests/hour with the BCM.

Automated operation and reliable technology

  • Increased throughput with a sample capacity of up to 350 colorimetric tests/hour and approximately 8 bitterness tests/hour.
  • Integrated platform simultaneously automates colorimetric tests, including beta-glucan, SO2, and pH, from the same sample.
  • Continuous access to samples, reagents, and cuvettes without interruption of routine workflow.
  • All necessary analysis steps are automated, providing true walk-away time for the operator.
  • Dilutions and repeat analyses are handled automatically.
  • Minimal sample pretreatment required.

Robust capability

  • Automated processes allow beer bittering substances to be extracted from interfering compounds found in the sample matrix prior to photometric analysis at 275nm.
  • Instrument versatility: Sample disk for 6 racks, 9 or 18 sample racks, 60 blocks for automated bitterness sample pretreatment, and 6 racks for bitterness reagents are configured.
  • Wide concentration range measurement, with excellent reproducibility.
  • User-specific applications can be developed to meet individual testing needs.
  • Over 20 system applications for beer and wort analysis.
  • Sensitivity: Ability to detect very low levels (ppb) of analytes.

Compact design and economical

  • System reagents eliminate time-consuming reagent preparation, freeing operators for other tasks.
  • Unique low volume cuvette design for less reagent waster and potentially lower operation costs.
  • Contamination-free analysis is achieved using disposable cuvettes.
  • Self-contained benchtop for small footprint.
  • No need for an external source of water.

Versatile and user-friendly

  • Open system – modify existing method or create your own test.
  • Accommodates a variety of sample cups and primary tubes.
  • No risk of carry-over or requirement to wash glassware.
  • Sample identification is automated by a barcode reader.
  • Automatic startup and shutdown protocols.
  • Intuitive graphic user-interface.
Specifications
Analytical MethodAutomating Labor-Intensive Testing, Bitterness Testing, Discrete Photometric Analyzer
CalibrationFactor, Bias, Linear, Logit-Log, Spline, Polynomial and Point-to-Point
CuvettesDiscrete Disposable Cuvettes. On Board Capacity 360 Measurement Cells.
Depth (English)27.5 in.
Depth (Metric)70 cm
DescriptionGallery Plus Beermaster Discrete Analyzer (with BCM Unit)
Distilled Water ConsumptionDeionized Water Consumption 2.5 L/hr.
Frequency50/60 Hz +/-5%
Height (English)24.4/51.1 in. (Cover Closed/Cover Open)
Height (Metric)62/130 cm (Cover Closed/Cover Open)
Measurement MethodPhotometric Analysis, Colorimetric
Operating SystemAutomated Photometric (Colorimetric and Enzymatic) Analysis Capable of Bitterness (BCM) Measurement
Quality Control DetailsReal-Time QC Program with Multiple and Variable Rules. Control Frequency User-Definable. Out-of-Specification Control Results Flagged.
QuantityEach
Reagents9-Position Sample Racks, Max 54 Positions (= 6 Racks) on-Board; Reagent Capacity 42 Positions on-Board
Sample DilutionAutomatic Sample Pre-Dilution. Automatic Sample Dilution of Over-Range Tests with Automatic re-Run. Addition of Manual Pre-Dilution Value for Result Calculation.
ThroughputUp to 350 Tests/hr.
Voltage100/240 V +/-10%
Weight (Metric)110 kg
Width (English)37 in.
Width (Metric)94 cm
Sample TypeFood and Beverage Sample, Environmental Sample
TypePhotometric Analyzer
Unit SizeEach

Frequently asked questions (FAQs)

What does the Bitterness Column Module (BCM) do in the Gallery Plus Beermaster discrete analyzer? What manual steps remain?

The BCM includes a coated extraction column, it automatically extracts bittering compounds from the beer matrix and then measures them photometrically at 275 nm. Manual sample prep is minimal (e.g., degassing by shaking). This module replaces the labor intensive manual/semi automated bitterness workflows found in many labs.

What are sample/reagent capacities and walk away time for the Gallery Plus Beermaster?

  • Sample disk: up to 6 racks, 9 or 18 position racks can be used (configuration can be affected when BCM hardware is installed).
  • Bitterness pretreatment: dedicated 60 position block plus a 6 position rack for bitterness reagents.
  • Reagent disk: 42 positions (cooled).
  • Cuvettes: 360 measurement cells on board.
  • Walk away time: typically 1–3 hours, depending on workload.

How fast is the Gallery Plus Beermaster analyzer?

  • Up to ~350 colorimetric tests/hour on the Gallery Plus Beermaster platform.
  • For bitterness via BCM, ~8 tests/hour is typical.
  • The bitterness workflow yields the first result in ~10 min, then about every 7 min thereafter.

Which parameters can the Gallery Plus Beermaster measure for beer/wort/malt?

The Gallery Plus Beermaster system can measure a variety of important parameters through the beer production process, from the source water to the final product. Critical parameters that the Gallery Plus Beermaster system can measure include: Bitterness (using the BCM), β-glucan, diastatic power (DP), alpha amylase (AA), FAN (NOPA), SO₂, color, pH, total protein, total polyphenols, sugars (glucose, fructose, sucrose), organic acids (lactic, malic, citric), ethanol, calcium, magnesium, iron, and more. Methods are optimized to ASBC/EBC/MEBAK reference result levels. The Gallery platform is a certified platform to run these applications per ASBC protocol. Specific Gallery test parameters can be found and downloaded from the ASBC website.

How is Gallery Plus Beermaster different from other beer analysis approaches (manual, semi automated, or flow/segmented flow analyzers)?

  • Automated bitterness: The Bitterness Column Module (BCM) extracts bittering substances from interfering matrix components and measures them at 275 nm, sharply reducing manual steps. It typically produces the first result in ~10 min and then about every 7 min in multi sample workflows.
  • Walk away discrete analysis: Discrete cell technology automates all steps and delivers results in minutes with minimal hands on time, Our discrete analyzer systems require lower sample/reagent volumes and offer easier operation than continuous flow methods.
  • Lower reagent use/waste: The platform’s low volume cuvettes and ready to use reagents are designed to cut reagent consumption and waste versus manual methods.