Indiko™ Ultra Clinical Chemistry Analyzer
Indiko™ Ultra Clinical Chemistry Analyzer
Indiko™ Ultra Clinical Chemistry Analyzer
Thermo Scientific™

Indiko™ Ultra Clinical Chemistry Analyzer

Indiko Ultra analyzers combine analytical credibility, operational clarity, advanced data protection, and scalable capacity in one dependable clinical chemistry analyzer portfolio.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCapacidad de procesamientoCapacidad
98741000-0Up to 350 photometric tests/h; up to 135 ISE tests/hUp to 108 samples with 18-position racks; up to 54 samples with 9-position racks
98730000-0Up to 200 photometric tests/hUp to 90 samples with 18-position racks; up to 45 samples with 9-position racks, 6-position reagents racks
98731000-0Up to 200 photometric tests/h; up to 120 ISE tests/hUp to 90 samples with 18-position racks; up to 45 samples with 9-position racks, 6-position reagents racks
98740000-0Up to 350 photometric tests/hUp to 108 samples with 18-position racks; up to 54 samples with 9-position racks
Número de catálogo 98741000-0
Precio (CLP)
-
Capacidad de procesamiento:
Up to 350 photometric tests/h; up to 135 ISE tests/h
Capacidad:
Up to 108 samples with 18-position racks; up to 54 samples with 9-position racks
The Thermo Scientific™ Indiko™ Ultra portfolio includes four models—Indiko Ultra, Indiko Ultra ISE, Indiko Ultra XL, and Indiko Ultra XL ISE—designed to meet diverse laboratory needs. Simplify daily operations with intuitive software, automated safeguards, and clear system visibility for consistent workflows. The compact benchtop design fits easily into your lab while helping to minimize resource use. Expand capabilities with flexible testing solutions that grow with your laboratory demands.
"
  • Everyday reliability
    • Enhanced cybersecurity architecture
    • Intuitive software with clear system status visibility
    • Automated clot detection and system checks
    • Maintenance-minimizing xenon light source
    • Direct ISE technology—no dilution required
    • Only one recommended preventive maintenance visit per year
  • Space-fitting solution
    • Compact footprint for space-constrained laboratory layouts
    • Low water consumption (~1.5 to 2.5 L/hour) reduces infrastructure requirements
    • Flexible waste handling options, including drain connectivity where applicable
  • Flexible capacity and performance
    • Optimized throughput for small to medium-sized laboratories
    • Continuous sample loading with up to 3 hours walkaway time
    • Integrated clinical chemistry and specialty testing support
  • "
    Especificaciones
    CapacidadUp to 108 samples with 18-position racks; up to 54 samples with 9-position racks
    Certificaciones/conformidadIVDR compliant
    CubetasYes, TENCELL cuvettes
    Dimensiones (Pr x An x Al)73 cm x 80 cm x 66/116 cm (with open cover)
    Tipo ISEDirect
    Temperatura de incubaciónControlled at 37°C
    Capacidad de procesamientoUp to 350 photometric tests/h; up to 135 ISE tests/h
    TipoClinical Chemistry Analyzer
    Unit SizeEach