Forma&trade; CO<sub>2</sub> Backup Systems
Fisher Scientific™
Forma™ CO2 Backup Systems
Receive the convenience, reassurance and reliability required in the event of a power or mechanical failure with this CO2 backup system.
Have Questions?
Número do catálogo 991505
Preço (BRL)
-
Obtain the convenience, reassurance and reliability required in the event of a power or mechanical failure with the Thermo Scientific™ Forma™ CO2 Backup Systems for ultra low temperature freezers, a must for critical storage applications.
Key Features:
  • Temperature is adjustable in 1°C increments to -75°C
  • Digital display indicates actual cabinet temperature
  • Indicator lights illuminate when system is activated
  • Liquid flow shuts off automatically when freezer door or lid is opened
  • System powered by rechargeable battery
  • Low-battery light indicates when battery requires charging
  • Additional indicator lights when cylinder is empty
Ordering Information:
Some models require freestanding system; others accept built-in systems. Specify freezer model and voltage when ordering.
Includes:
All necessary hardware except CO2 cylinder (must be ordered separately)
Especificações
DefrostAutomatic
DescriptionHigh-pressure Hose
StyleBuilt-in
Temperature Range-75°C
Unit SizeEach